Orders LinkContactUsLinkComponent
VWR®, 使い捨て計量ボート
VWR®, 使い捨て計量ボート
Supplier:  VWR Collection
Certificates
VWR®, 使い捨て計量ボート
VWR®, 使い捨て計量ボート
25 Options Available Below
Product Details & Documents
PS, square.
  • Shallow form with wide, flat bottom avoids tipping
  • Rounded corners simplify transfer
  • Temperature resistant up to 70 °C
These flexible weighing boats, with smooth surfaces, enable accurate pour-out with minimal sample loss. They are biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bleaches. Ideal for use with microwaves, balances or moisture analysers.
 : These weighing boats are produced and packed in plastic bags in a cleanroom.
Product Family Options
Standard
Product Information
  • 容量
    W×D×H
    Colour
    Availability
    Price
  • 7 ml
    46×46×8 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 7 ml
    46×46×8 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 7 ml
    46×46×8 mm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 7 ml
    46×46×8 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $41.70
    $41.70
    1 * 500 items
       $41.70
  • 7 ml
    46×46×8 mm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $44.39
    $44.39
    1 * 500 items
       $44.39
  • 100 ml
    85×85×24 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 100 ml
    85×85×24 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 100 ml
    85×85×24 mm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 100 ml
    85×85×24 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $68.32
    $68.32
    1 * 500 items
       $68.32
  • 250 ml
    140×140×22 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 250 ml
    140×140×22 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 250 ml
    140×140×22 mm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 250 ml
    140×140×22 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $151.87
    $151.87
    1 * 500 items
       $151.87
  • 250 ml
    140×140×22 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $165.18
    $165.18
    1 * 500 items
       $165.18
Anti-static
Product Information
  • 容量
    W×D×H
    Colour
    Availability
    Price
  • 7 ml
    46×46×8 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 7 ml
    46×46×8 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 7 ml
    46×46×8 mm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 100 ml
    85×85×24 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 100 ml
    85×85×24 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $67.38
    $67.38
    1 * 500 items
       $67.38
  • 100 ml
    85×85×24 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 100 ml
    85×85×24 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - $66.99
    $66.99
    1 * 500 items
       $66.99
  • 100 ml
    85×85×24 mm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 250 ml
    140×140×22 mm
    White
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 250 ml
    140×140×22 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 250 ml
    140×140×22 mm
    Black