Orders LinkContactUsLinkComponent
VWR®, ビーカー、低形状
Supplier:   VWR Collection
VWR®, ビーカー、低形状
Supplier:  VWR Collection
Certificates
VWR®, ビーカー、低形状
VWR®, ビーカー、低形状
14 Options Available Below
Product Details & Documents
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.
Product Family Options
Product Information
  • 容量
    高さ
    外径
    Availability
    Price
  • 1 ml
    18 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 5 ml
    26 mm
    20 mm
    1 * 1 items
  • 10 ml
    33 mm
    24 mm
    1 * 1 items
  • 25 ml
    47 mm
    32 mm
    1 * 1 items
  • 50 ml
    60 mm
    43 mm
    1 * 1 items
  • 100 ml
    68 mm
    54 mm
    1 * 1 items
  • 150 ml
    69 mm
    59 mm
    1 * 1 items
  • 250 ml
    97 mm
    66 mm
    1 * 1 items
  • 400 ml
    106 mm
    80 mm
    1 * 1 items
  • 500 ml
    125 mm
    80 mm
    1 * 1 items
  • 600 ml
    125 mm
    90 mm
    1 * 1 items
  • 1000 ml
    155 mm
    100 mm
    1 * 1 items
  • 2000 ml
    210 mm
    120 mm
    1 * 1 items
  • 5000 ml
    280 mm
    170 mm