Orders LinkContactUsLinkComponent
VWR®, バッグ、バイオハザード
Supplier:   VWR Collection
VWR®, バッグ、バイオハザード
Supplier:  VWR Collection
Certificates
VWR®, バッグ、バイオハザード
VWR®, バッグ、バイオハザード
2 Options Available Below
Product Details & Documents
Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 75 µm thick, orange, with 'Biohazard' label.
  • Autoclavable up to 135 °C
  • The useful temperature indicator text is white before autoclaving, then black
  • 'Contents Have Been Autoclaved' indicates successful autoclaving
  • Luminous red for easy recognition and highly visible 'Biohazard' label with the corresponding warning sign
These bags are DOT approved.
Product Family Options
Product Information
  • Material
    容量
    長さ×幅
    厚さ
    Availability
    Price
  • PP
    NA
    762×610 mm
    75 µm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - $895.94
    $895.94
    1 * 250 items
       $895.94
  • PP
    60 L
    914×610 mm
    100 µm
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - $1,602.99
    $1,602.99
    1 * 200 items
       $1,602.99