VWR® Conductivity standard, CRM, 5000 μS/cm
85645.260
:
- パックタイプ:Plastic bottle
- Pk:500 mL
- 説明:KCl 0,0375 mol/l: 5000 µS/cm
- Seq:26
Directly traceable to NIST, tested and certified to ISO17025 and ISO 17034
Directly traceable to NIST, tested and certified to ISO17025 and ISO 17034