ContactUsLinkComponent
VWR® Sterile Coveralls, 10⁻⁶ SAL
VWR® Sterile Coveralls, 10⁻⁶ SAL
Supplier:  VWR Collection
New Product
Certificates
undefined
VWR® Sterile Coveralls, 10⁻⁶ SAL
Supplier:  VWR Collection
New Product
Specifications
  • Catalog No:
  • VWRU89523-466
  • VWRU89523-464
  • Garment application:
  • Pharmaceutical, Electronics/semi-conductor
  • Pharmaceutical, Electronics/semi-conductor
  • Bacterial filtration efficiency:
  • ≥90%
  • ≥90%