VWR®, マルチウェル細胞培養プレート
VWR® multiwell cell culture plates are available with a choice of surface treatments.
- Uniform well volume ensures an equal growth surface area
- Well surface is smooth and free from striations to maximise usable growth area
- Raised rims on wells with uniform rings on the lid to reduce evaporation
- Single-position lid reduces the risk of cross-contamination and handling mistakes
- Wells are labelled with alphanumeric code for easy identification
- Suitable for use with all common instruments and automation
- Sterilised by electron beam irradiation
- Free from DNase and RNase, and non pyrogenic
Increased cell attachment surface treatment: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
TC-treated: These plates are vacuum-gas plasma-treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture plates are ideal for applications where cell attachment is not desired.
Culture area = cm² per well.
Certifications: ISO 13485:2003, ISO 9001:2008
Packaging: Individually wrapped in peel-to-open paper/plastic blister packs.
VWR® multiwell cell culture plates are available with a choice of surface treatments.
- Uniform well volume ensures an equal growth surface area
- Well surface is smooth and free from striations to maximise usable growth area
- Raised rims on wells with uniform rings on the lid to reduce evaporation
- Single-position lid reduces the risk of cross-contamination and handling mistakes
- Wells are labelled with alphanumeric code for easy identification
- Suitable for use with all common instruments and automation
- Sterilised by electron beam irradiation
- Free from DNase and RNase, and non pyrogenic
Increased cell attachment surface treatment: This highly hydrophilic surface offers a significant advantage over the traditional cell culture surface. The surface treatment can improve cell spreading and attachment, and is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
TC-treated: These plates are vacuum-gas plasma-treated for consistent cell attachment and growth.
Non-treated: Non-treated cell and tissue culture plates are ideal for applications where cell attachment is not desired.
Culture area = cm² per well.
Certifications: ISO 13485:2003, ISO 9001:2008
Packaging: Individually wrapped in peel-to-open paper/plastic blister packs.
Documents
Avantor Cell Culture Solutions.pdf
Product Literature
CoC VWR Multi Well Cell Culture Plates Standard Line CoC VWR Multiwell Cell Culture Plates CoC VWR Multiwell Cell Culture Plates COC_VWR_CELL_ATTACHMENT_TISSUE_CULTURE_PLATE_TREATED COC_VWR_CELL_ATTACHMENT_TISSUE_CULTURE_PLATES_FLAT_BOTTOM_TREATED
CoC VWR® Multiwell Cell Culture Plates.pdf
Conformity declaration
- Catalog No:
- 732-2905
- 732-2904
- 734-2798
- 734-2323
- 734-2777
- 734-2799
- 734-2324
- 734-2778
- 734-2800
- 734-2325
- 734-2779
- 734-2801
- 734-2326
- 734-2780
- 734-2888
- 734-2802
- 734-2327
- 734-2781
- 734-2803
- 734-2328
- 734-2782
- 732-2907
- 732-2906
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Product Family Options
- 説明ColourCulture areaRecommended working volumeSterilePacked
Low Manufacturing Impact Specifications
Cat. No.732-2905Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.732-2904Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 6-well plates, flat bottom, treated for increased cell attachmentClear9,6 cm²1,9 - 2,9 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2798Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.734-2323Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact Specifications
Cat. No.734-2777Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M- 12-well plates, flat bottom, treated for increased cell attachmentClear3,85 cm²0,76 - 1,14 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2799Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M Low Manufacturing Impact Specifications
Cat. No.734-2324Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 12-well plates, flat bottom, non-treatedClear3,85 cm²0,76 - 1,14 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2778Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 24-well plates, flat bottom, treated for increased cell attachmentClear1,93 cm²0,38 - 0,57 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2800Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M Low Manufacturing Impact Specifications
Cat. No.734-2325Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M- 24-well plates, flat bottom, non-treatedClear1,93 cm²0,38 - 0,57 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2779Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 48-well plates, flat bottom, treated for increased cell attachmentClear0,83 cm²0,19 - 0,29 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2801Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.734-2326Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M- 48-well plates, flat bottom, non-treatedClear0,83 cm²0,19 - 0,29 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2780Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 48-well plates, flat bottom, non treatedClear0,83 cm²0,19 - 0,29 ml-1/bag1 * 200 items
Low Manufacturing Impact Specifications
Cat. No.734-2888Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 96-well plates, flat bottom, treated for increased cell attachmentClear0,33 cm²0,075 - 0,20 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2802Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 96-well plates, flat bottom, TC-treatedClear0,33 cm²0,075 - 0,20 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2327Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 96-well plates, flat bottom, non-treatedClear0,33 cm²0,075 - 0,20 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2781Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 96-well plates, round bottom, treated for increased cell attachmentClear0,58 cm²0,075 - 0,20 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2803Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 96-well plates, round bottom, TC-treatedClear0,58 cm²0,075 - 0,20 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2328Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M - 96-well plates, round bottom, non-treatedClear0,58 cm²0,075 - 0,20 ml+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.734-2782Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0M Low Manufacturing Impact Specifications
Cat. No.732-2907Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 384-well plates, flat bottom, non-treatedClear0,1135 cm²10 - 100 μl+1/bag1 * 100 items
Low Manufacturing Impact Specifications
Cat. No.732-2906Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
Breatheable rayon film seals for biological cultures
- Sterile説明厚さ光学特性