Order Entry
Japan
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™
Irritant
EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR2580-12
CAS Number:  
Irritant
EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR2580-12
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • ACS - Assay (C₁₀H₁₆N₂O₈):
    99.4 - 100.6 %
  • ACS - Calcium (Ca):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Insoluble in Dilute NH₄OH:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤5 ppm
  • ACS - Nitriloacetic Acid:
    ≤0.1 %
  • ACS - Residue after Ignition:
    ≤0.2 %

Specification Test Results

About this item

 :
 :292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0,86 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
 :GHS07
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541