Orders LinkContactUsLinkComponent
VWR® Sample Vials
VWR® Sample Vials
VWR® Sample Vials
Supplier:  VWR Collection
Certificates
VWR® Sample Vials
VWR® Sample Vials
7 Options Available Below
Product Details & Documents
Screw neck vials with flat bottom made from clear borosilicate glass.
  • Highly resistant against chemicals
  • High temperature resistance
  • Delivered with screw caps
Ideal for storage of liquid and powders.
Product Family Options
Product Information
  • 容量
    Base
    Ø×H
    Colour
    キャップ付き
    キャップサイズ
    Cap material
    Cap colour
    隔膜
    Sterile
    Availability
    Price
  • 2,00 ml
    Flat bottom
    12×35 mm
    Clear
    Yes
    8-425
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 4,00 ml
    Flat bottom
    15×45 mm
    Clear
    Yes
    13-425
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 6,00 ml
    Flat bottom
    16×50 mm
    Clear
    Yes
    13-425
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 8,00 ml
    Flat bottom
    17×60 mm
    Clear
    Yes
    15-425
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 12,00 ml
    Flat bottom
    19×65 mm
    Clear
    Yes
    15-425
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 16,00 ml
    Flat bottom
    21×70 mm
    Clear
    Yes
    18-400
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 24,00 ml
    Flat bottom
    23×85 mm
    Clear
    Yes
    20-400
    PP
    Black
    PTFE liner
    -
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items