Orders LinkContactUsLinkComponent
VWR® Serum/Sample Vials
VWR® Serum/Sample Vials
VWR® Serum/Sample Vials
Supplier:  VWR Collection
Certificates
VWR® Serum/Sample Vials
VWR® Serum/Sample Vials
5 Options Available Below
Product Details & Documents
Crimp neck vials with flat bottom made from clear borosilicate glass.
  • Highly resistant against chemicals
  • High temperature resistance
  • Can be used as vaccine or injectable drug containers
  • Stoppers and caps have to be ordered separately
Ideal for long-term sample storage or lyophilisation.
Product Family Options
Product Information
  • 説明
    容量
    Base
    Ø×H
    Colour
    キャップ付き
    Sterile
    Availability
    Price
  • Neck size: ND11
    2,00 ml
    Flat bottom
    10,25×45,00 mm
    Clear
    No
    -
    Log in for availability
    Log in for availability
    1 * 270 items
    1 * 270 items
  • Neck size: ND13
    3,00 ml
    Flat bottom
    16,25×35,00 mm
    Clear
    No
    -
    Log in for availability
    Log in for availability
    1 * 209 items
    1 * 209 items
  • Neck size: ND20
    5,00 ml
    Flat bottom
    24,00×33,00 mm
    Clear
    No
    -
    Log in for availability
    Log in for availability
    1 * 91 items
    1 * 91 items
  • Neck size: ND20
    20,00 ml
    Flat bottom
    24,00×74,00 mm
    Clear
    No
    -
    Log in for availability
    Log in for availability
    1 * 145 items
    1 * 145 items
  • Neck size: ND20
    30,00 ml
    Flat bottom
    30,00×62,00 mm
    Clear
    No
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items