Orders LinkContactUsLinkComponent
Avantor® Rigid Outer Support Containers
Avantor® Rigid Outer Support Containers
Supplier:   VWR Collection
Avantor® Rigid Outer Support Containers
Supplier:  VWR Collection
Certificates
Avantor® Rigid Outer Support Containers
Avantor® Rigid Outer Support Containers
12 Options Available Below
Product Details & Documents
3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropelene (PP), or stainless steel material.
  • Standard 3D cubic bags are available in 100, 200, and 1000 L volumes with 3 top ports
  • Standard 3D cubic bags are available with or without a bottom port
  • Cleanroom and shipping variants are available in either material format
  • Plastic containers can collapse for easy storage
  • Stainless steel containers are ISTA certified for shipping
Avantor® 3D cubic bags are made from a proprietary film which features a polyethelene inner and outer layer with gas and moisture barrier layer between. The film has high capacity and flexibility, and is resistant to a wide range of chemicals.
Certifications: Stainless steel containers meet ISTA 3,1B certification.
Product Family Options
Plastic, foldable, cleanroom version
Product Information
  • 説明
    容量
    Material
    Availability
    Price
  • Rigid outer support container, foldable, cleanroom version
    250 L
    Plastic
    1 * 1 items
  • Rigid outer support container, foldable, cleanroom version
    500 L
    Plastic
    1 * 1 items
  • Rigid outer support container, foldable, cleanroom version
    1000 L
    Plastic
    1 * 1 items
Plastic, foldable, transport version
Product Information
  • 説明
    容量
    Material
    Availability
    Price
  • Rigid outer support container, foldable, transport version
    250 L
    Plastic
    1 * 1 items
  • Rigid outer support container, foldable, transport version
    500 L
    Plastic
    1 * 1 items
  • Rigid outer support container, foldable, transport version
    1000 L
    Plastic
    1 * 1 items
Stainless steel, cleanroom version
Product Information
  • 説明
    容量
    Material
    Availability
    Price
  • Rigid outer support container, cleanroom version
    200 L
    Stainless steel
    1 * 1 items
  • Rigid outer support container, cleanroom version
    500 L
    Stainless steel
    1 * 1 items
  • Rigid outer support container, cleanroom version
    1000 L
    Stainless steel
    1 * 1 items
Stainless steel, transport version
Product Information
  • 説明
    容量
    Material
    Availability
    Price
  • Rigid outer support container, transport version
    200 L
    Stainless steel
    1 * 1 items
  • Rigid outer support container, transport version
    500 L
    Stainless steel
    1 * 1 items
  • Rigid outer support container, transport version
    1000 L
    Stainless steel
    1 * 1 items
Complementary Products
Product Information
  • Catalog #
    説明
    容量
    Availability
    Price
  • 734-2961
    3D cubic bags with 2× 50 cm silicone platinum ports
    Port 1: 1,27×1,90 mm, with pinch clamp and 1× male MPX
    Port 2: 1,27×1,90 mm, with pinch clamp and 1× female MPX
    500 l
    Log in for availability
    1 * 1 items - $0.00
  • 734-2960
    3D cubic bags with 2× 50 cm silicone platinum ports
    Port 1: 1,27×1,90 mm, with pinch clamp and 1× male MPX
    Port 2: 1,27×1,90 mm, with pinch clamp and 1× female MPX
    200 l
    Log in for availability
    1 * 5 items - $0.00