Smiths Medical® Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 200/PK
MFLX30526-34BLK
- Flow pattern:Three-Way
- Max. pressure:1050 psi
- 説明:Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 200/PK
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Pk:200
- Inlet port:Female Luer(2)
- Outlet port:Male Luer Lock
Pressure rating up to 1050 psi.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Stopcock fittings feature luer connections.
- Flow pattern:Three-Way
- Max. pressure:1050 psi
- 説明:Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 200/PK
- Environmentally Preferable:
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Pk:200
- Inlet port:Female Luer(2)
- Outlet port:Male Luer Lock