Orders LinkContactUsLinkComponent
Copper sulphate solution
Copper sulphate solution
Plastic bottle, 5 L
Catalog # 87204.360
Supplier:   VWR Chemicals
Copper sulphate solution
Catalog # 87204.360
Supplier:  VWR Chemicals
CAS Number:  7758-99-8
Copper sulphate solution
Copper sulphate solution
Plastic bottle, 5 L
CAS Number:  7758-99-8
Price
$371.20
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Copper(II) sulphate, Cupric sulphate
  • CAS Number:7758-99-8
  • Formula:CuSO₄·5H₂O
  • MDL Number:MFCD00149681
  • Molecular Weight:249.69 g/mol
  • Storage Temperature:Ambient
Specifications
Specification Test Results
  • Density:
    1.165 - 1.195
Specifications
  • パックタイプ:
    Plastic bottle
  • Pk:
    5 L
Specifications