Palladium matrix modifier 5 g/L in nitric acid 1% [99,999% Pd(NO₃)₂], ARISTAR® for graphite furnace AAS
Catalog # 91281.154
Supplier: VWR Chemicals
CAS Number:
Specifications
- パックタイプ:Plastic bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS07