VWR®, 基本保護用ディスポーザブルフード
Catalog # 112-6248
Supplier: VWR Collection
Specifications
- 衣服のサイズ:L
- Garment application:Laboratory
- Colour:White
- 説明:VWR® Basic protection hood
- Pk:500
Specifications
About this item
Disposable non-woven hood is made of 20 gsm SPP fabric, and features a small open face hole.
- Full garment dimensions are 45,7W×50,8L cm (18×20")
- Constructed from 20 gsm SPP fabric
Hoods provide reliable protection in laboratory, electrical, pharmaceutical, food, industrial, and cleanroom applications.
Certifications: ISO9001.