Masterflex® Console Gear Pump Drive for MicroPump A-Mount Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Simple variable speed control handles many fluid handling applications.
- 100:1 speed-control range controls flow over a wide range
- Maximum continuous-duty flow rates possible with Micropump® A-mount pump heads
- Simple operation with single-turn speed control
- Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
- Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Couple with an A-mount gear pump head for pulseless fluid flow.
Ordering information: Pump head is not included.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX75211-17-EU
- MFLX75211-17-DK
- Duty cycle continuous:
- Yes
- Yes
- Enclosure IP rating:
- IP 22
- IP 22
- Min. flow rate:
- 0,85 ml/min
- 0,85 ml/min
- Min. motor RPM:
- 50 rpm
- 50 rpm
- Motor enclosure type:
- Painted steel and ABS plastic
- Painted steel and ABS plastic
- Motor hp:
- 1/20
- 1/20
- Remote control input:
- Start/stop
- Start/stop
- Warranty:
- 2 years
- 2 years