マイクロポンプ® B-マウントギアポンプヘッド
About this item
Smooth pulseless flow provides precise, leak-free delivery of fluids.
- Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
- Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.
15 Options Available Below
Product Details & Documents
Smooth pulseless flow provides precise, leak-free delivery of fluids.
- Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
- Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX74004-01
- MFLX74004-05
- MFLX74004-07
- MFLX74004-09
- MFLX74004-11
- MFLX74004-17
- MFLX74004-23
- MFLX74004-27
- MFLX74004-29
- MFLX74004-31
- MFLX74004-33
- MFLX74004-71
- MFLX74004-73
- MFLX74004-75
- MFLX74004-03
- Body material:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Differential pressure:
- 3,5 bar (50 psi)
- 5,2 bar (75 psi)
- 3,5 bar (50 psi)
- 3,5 bar (50 psi)
- 3,5 bar (50 psi)
- 5,2 bar (75 psi)
- 2,8 bar (40 psi)
- 5,2 bar (75 psi)
- 2,8 bar (40 psi)
- 2,8 bar (40 psi)
- 3,5 bar (50 psi)
- 5,2 bar (75 psi)
- 5,2 bar (75 psi)
- 5,5 bar (80 psi)
- 3,5 bar (50 psi)
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Gear material:
- PTFE
- PPS
- PTFE
- PTFE
- PTFE
- PPS
- Carbon graphite
- PPS
- Carbon graphite
- Carbon graphite
- PTFE
- PEEK
- PEEK
- PEEK
- PTFE
- Inlet connection:
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- Internal bypass:
- Yes
- Yes
- No
- No
- No
- No
- No
- No
- No
- No
- Yes
- No
- No
- No
- Yes
- Max. temperature:
- 54 °C (130 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 54 °C (130 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- Max. viscosity:
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- Min. temperature:
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- Min. viscosity:
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- 0,5 cP
- Outlet connection:
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- 0,125 NPT(F)
- Particulates:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Seal material:
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- Series:
- GJ
- GJ
- GJ
- GJ
- GJ
- GJ
- GA
- GA
- GA
- GA
- GJ
- GA
- GA
- GJ
- GJ
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifications
Product Family Options
Product Information
- 説明流量Max. motor RPMAvailabilityPrice
- Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0,64 ml/rev0,64 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-01Body materialStainless steelDifferential pressure3,5 bar (50 psi)Duty cycle continuousYesGear materialPTFEInlet connection0,125 NPT(F)Internal bypassYesMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PPS Gear, PTFE Seal, 0,64 ml/rev0,64 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-05Body materialStainless steelDifferential pressure5,2 bar (75 psi)Duty cycle continuousYesGear materialPPSInlet connection0,125 NPT(F)Internal bypassYesMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0,316 ml/rev0,316 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-07Body materialStainless steelDifferential pressure3,5 bar (50 psi)Duty cycle continuousYesGear materialPTFEInlet connection0,125 NPT(F)Internal bypassNoMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0,64 ml/rev0,64 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-09Body materialStainless steelDifferential pressure3,5 bar (50 psi)Duty cycle continuousYesGear materialPTFEInlet connection0,125 NPT(F)Internal bypassNoMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0,91 ml/rev0,91 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-11Body materialStainless steelDifferential pressure3,5 bar (50 psi)Duty cycle continuousYesGear materialPTFEInlet connection0,125 NPT(F)Internal bypassNoMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PPS Gear, PTFE Seal, 0,64 ml/rev0,64 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-17Body materialStainless steelDifferential pressure5,2 bar (75 psi)Duty cycle continuousYesGear materialPPSInlet connection0,125 NPT(F)Internal bypassNoMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Suction Shoe Style, Stainless Steel, Carbon Graphite Gear, PTFE Seal, 0,017 ml/rev0,017 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-23Body materialStainless steelDifferential pressure2,8 bar (40 psi)Duty cycle continuousYesGear materialCarbon graphiteInlet connection0,125 NPT(F)Internal bypassNoMax. temperature177 °C (350 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGAWarranty1 year - Gear Pump Head, Suction Shoe Style, Stainless Steel, PPS Gear, PTFE Seal, 0,017 ml/rev0,017 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-27Body materialStainless steelDifferential pressure5,2 bar (75 psi)Duty cycle continuousYesGear materialPPSInlet connection0,125 NPT(F)Internal bypassNoMax. temperature177 °C (350 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGAWarranty1 year - Gear Pump Head, Suction Shoe Style, Stainless Steel, Carbon Graphite Gear, PTFE Seal, 0,042 ml/rev0,042 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-29Body materialStainless steelDifferential pressure2,8 bar (40 psi)Duty cycle continuousYesGear materialCarbon graphiteInlet connection0,125 NPT(F)Internal bypassNoMax. temperature177 °C (350 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGAWarranty1 year - Gear Pump Head, Suction Shoe Style, Stainless Steel, Carbon Graphite Gear, PTFE Seal, 0,084 ml/rev0,084 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-31Body materialStainless steelDifferential pressure2,8 bar (40 psi)Duty cycle continuousYesGear materialCarbon graphiteInlet connection0,125 NPT(F)Internal bypassNoMax. temperature177 °C (350 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGAWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0,64 ml/rev0,64 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-33Body materialStainless steelDifferential pressure3,5 bar (50 psi)Duty cycle continuousYesGear materialPTFEInlet connection0,125 NPT(F)Internal bypassYesMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Suction Shoe Style, Stainless Steel, PEEK Gear, PTFE Seal, 0,017 ml/rev0,017 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-71Body materialStainless steelDifferential pressure5,2 bar (75 psi)Duty cycle continuousYesGear materialPEEKInlet connection0,125 NPT(F)Internal bypassNoMax. temperature177 °C (350 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGAWarranty1 year - Gear Pump Head, Suction Shoe Style, Stainless Steel, PEEK Gear, PTFE Seal, 0,092 ml/rev0,092 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-73Body materialStainless steelDifferential pressure5,2 bar (75 psi)Duty cycle continuousYesGear materialPEEKInlet connection0,125 NPT(F)Internal bypassNoMax. temperature177 °C (350 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGAWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PEEK Gear, PTFE Seal, 0,64 ml/rev0,64 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-75Body materialStainless steelDifferential pressure5,5 bar (80 psi)Duty cycle continuousYesGear materialPEEKInlet connection0,125 NPT(F)Internal bypassNoMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year - Gear Pump Head, Cavity Style, Stainless Steel, PTFE Gear, PTFE Seal, 0,91 ml/rev0,91 ml/rev5500 rpmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX74004-03Body materialStainless steelDifferential pressure3,5 bar (50 psi)Duty cycle continuousYesGear materialPTFEInlet connection0,125 NPT(F)Internal bypassYesMax. temperature54 °C (130 °F)Max. viscosity1500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,5 cPOutlet connection0,125 NPT(F)ParticulatesNoSeal materialPTFESeriesGJWarranty1 year