Masterflex® Printed Microscope Slide, eight well; 144/pk
Catalog # MFLX75955-27
Supplier: Avantor Fluid Handling
Specifications
- ウェルの数:8
- 説明:Microscope slides, printed with mask
- 長さ×幅:76,2×25,4 mm
- Pk:144
- Slide type:Printed with Mask
Specifications
About this item
Keep samples isolated.
- Screen-printed well design prevents bleed-through
- Hydrophobic epoxy ink withstands frequent washing and resists chemicals
- Contrasting ink makes orienting the microscope field of view over a sample easy
These printed microscope slides feature circular wells designed to hold samples in place, allowing multiple wells per slide, so you can increase throughput without adding to cleanup. Black PTFE-based epoxy makes locating wells with a microscope simple and quick, improving efficiency. Select from a range of well counts to suit any application.
- ウェルの数:8
- 説明:Microscope slides, printed with mask
- 長さ×幅:76,2×25,4 mm
- Environmentally Preferable:
- Pk:144
- Slide type:Printed with Mask