Orders LinkContactUsLinkComponent
DC電源式ミニダイヤフラムポンプ
DC電源式ミニダイヤフラムポンプ
DC電源式ミニダイヤフラムポンプ
DC電源式ミニダイヤフラムポンプ
DC-powered pumps with PVDF pump head.
  • High resolution with a flow range of 0.055 to 38 ml/min
  • Choose between 12 VDC or 24 VDC power—pulse, start/stop, or proportional control
  • Connect 1/8" x 1/4" tubing directly to pump ports
Choose a pump with one of the following control methods:
Pulse: 1 stroke per pulse—0 to 720 strokes per minute control capability
4 to 20 mA: proportional control based on mA signal
1 to 5 VDC: proportional control based upon a 1 to 5 VDC input signal
Start/stop: pump stops during contact input
  • Accuracy:
  • ±3%
  • ±3%
  • ±3%
  • ±3%
  • ±3%
  • Duty cycle intermittent:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • Input:
  • Pulse
  • 4-20 mA Proportional
  • 4-20 mA Proportional
  • 0-5 V Proportional
  • 0-5 V Proportional
  • Max. pressure:
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • Max. temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Max. viscosity:
  • 50 cP
  • 50 cP
  • 50 cP
  • 50 cP
  • 50 cP
  • Min. flow rate:
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • Outlet connection:
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • Wetted materials:
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • 説明
    最大流量
    電力
    高さ
    長さ
  • HRP54V-1P3 DC-Powered Diaphragm Pump; 12 VDC, Pulse
    38 ml/min (0,6 gal./h, 720 strokes/min)
    12 VDC; 1,5 A
    73 mm
    10,1 cm
    6,2 cm
    1 * 1 items
    1 * 1 items
    CE Compliant
    IP65 washdown
    Specifications
    Cat. No.
    MFLX74201-01
    Accuracy
    ±3%
    Duty cycle intermittent
    Yes
    Inlet connection
    1/8" ID x 1/4" OD compression
    Input
    Pulse
    Max. pressure
    29 bar (2 psi)
    Max. temperature
    40 °C (104 °F)
    Max. viscosity
    50 cP
    Min. flow rate
    0,055 ml/min (0,001 gal./h; 0 strokes/min)
    Outlet connection
    1/8" ID x 1/4" OD compression
    Wetted materials
    Ceramic, FKM, PTFE, PVDF
    Warranty
    1 year
    CE Compliant
    IP65 washdown
  • HRP54V-1A3 DC-Powered Diaphragm Pump; 12 VDC, 4-20 mA Proportional
    38 ml/min (0,6 gal./h, 720 strokes/min)
    12 VDC; 1,5 A
    73 mm
    10,1 cm
    6,2 cm
    1 * 1 items
    1 * 1 items
    CE Compliant
    IP65 washdown
    Specifications
    Cat. No.
    MFLX74201-11
    Accuracy
    ±3%
    Duty cycle intermittent
    Yes
    Inlet connection
    1/8" ID x 1/4" OD compression
    Input
    4-20 mA Proportional
    Max. pressure
    29 bar (2 psi)
    Max. temperature
    40 °C (104 °F)
    Max. viscosity
    50 cP
    Min. flow rate
    0,055 ml/min (0,001 gal./h; 0 strokes/min)
    Outlet connection
    1/8" ID x 1/4" OD compression
    Wetted materials
    Ceramic, FKM, PTFE, PVDF
    Warranty
    1 year
    CE Compliant
    IP65 washdown
  • HRP54V-2A3 DC-Powered Diaphragm Pump; 24 VDC, 4-20 mA Proportional
    38 ml/min (0,6 gal./h, 720 strokes/min)
    24 VDC; 1 A
    73 mm
    10,1 cm
    6,2 cm
    1 * 1 items
    1 * 1 items
    CE Compliant
    IP65 washdown
    Specifications
    Cat. No.
    MFLX74201-13
    Accuracy
    ±3%
    Duty cycle intermittent
    Yes
    Inlet connection
    1/8" ID x 1/4" OD compression
    Input
    4-20 mA Proportional
    Max. pressure
    29 bar (2 psi)
    Max. temperature
    40 °C (104 °F)
    Max. viscosity
    50 cP
    Min. flow rate
    0,055 ml/min (0,001 gal./h; 0 strokes/min)
    Outlet connection
    1/8" ID x 1/4" OD compression
    Wetted materials
    Ceramic, FKM, PTFE, PVDF
    Warranty
    1 year
    CE Compliant
    IP65 washdown
  • HRP54V-1V3 DC-Powered Diaphragm Pump; 12 VDC, 0-5 V Proportional
    38 ml/min (0,6 gal./h, 720 strokes/min)
    12 VDC; 1,5 A
    73 mm
    10,1 cm
    6,2 cm
    1 * 1 items
    1 * 1 items
    CE Compliant
    IP65 washdown
    Specifications
    Cat. No.
    MFLX74201-21
    Accuracy
    ±3%
    Duty cycle intermittent
    Yes
    Inlet connection
    1/8" ID x 1/4" OD compression
    Input
    0-5 V Proportional
    Max. pressure
    29 bar (2 psi)
    Max. temperature
    40 °C (104 °F)
    Max. viscosity
    50 cP
    Min. flow rate
    0,055 ml/min (0,001 gal./h; 0 strokes/min)
    Outlet connection
    1/8" ID x 1/4" OD compression
    Wetted materials
    Ceramic, FKM, PTFE, PVDF
    Warranty
    1 year
    CE Compliant
    IP65 washdown
  • HRP54V-2V3 DC-Powered Diaphragm Pump; 24 VDC, 0-5 V Proportional
    38 ml/min (0,6 gal./h, 720 strokes/min)
    24 VDC; 1 A
    73 mm
    10,1 cm
    6,2 cm
    1 * 1 items
    1 * 1 items
    CE Compliant
    IP65 washdown
    Specifications
    Cat. No.
    MFLX74201-23
    Accuracy
    ±3%
    Duty cycle intermittent
    Yes
    Inlet connection
    1/8" ID x 1/4" OD compression
    Input
    0-5 V Proportional
    Max. pressure
    29 bar (2 psi)
    Max. temperature
    40 °C (104 °F)
    Max. viscosity
    50 cP
    Min. flow rate
    0,055 ml/min (0,001 gal./h; 0 strokes/min)
    Outlet connection
    1/8" ID x 1/4" OD compression
    Wetted materials
    Ceramic, FKM, PTFE, PVDF
    Warranty
    1 year
    CE Compliant
    IP65 washdown