Orders LinkContactUsLinkComponent
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Certificates
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, Avantor®
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, Avantor®
6 Options Available Below
Product Details & Documents
Ideal for general transfer applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: General laboratory use
Characteristics: Nontoxic, nonaging, nonoxidizing, easy flow monitoring, transparent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Specifications for Product Family
Specifications
  • Model:
  • 06509-13
  • 06509-14
  • 06509-16
  • 06509-17
  • 06509-18
  • 06509-25
  • Length per pack:
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • Max. pressure:
  • 2,7 bar (40 psi)
  • 2,7 bar (40 psi)
  • 2,7 bar (40 psi)
  • 1,4 bar (20 psi)
  • 1 bar (15 psi)
  • 2,4 bar (35 psi)
  • Min. temperature:
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • –46 °C (–51 °F)
  • Max. temperature:
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • 74 °C (165 °F)
  • Min. temperature (dynamic):
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • Max. temperature (dynamic):
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Max. vacuum:
  • 660 mm Hg (26" Hg)
  • 660 mm Hg (26" Hg)
  • 660 mm Hg (26" Hg)
  • 510 mm Hg (20" Hg)
  • 510 mm Hg (20" Hg)
  • 660 mm Hg (26" Hg)
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Regulatory approvals:
  • ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
  • ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
  • ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
  • ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
  • ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
  • ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
  • Sterilisation:
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Autoclave, Ethylene Oxide
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • No
  • No
  • No
  • No
  • No
  • No
  • Suction lift H₂O:
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • 6,7 m (22 ft.)
  • 6,7 m (22 ft.)
  • 8,8 m (29 ft.)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • 説明
    チューブ内径
    Tubing material
    L/S Masterflex tubing size
    Hose barb size
    最大流量
    Availability
    Price
  • Tygon E-Lab (E-3603) Pump Tubing, L/S 13, 50 ft.
    0,80 mm (0,031")
    Tygon® E-Lab (E-3603)
    13
    0,0625"
    36 ml/min
    Log in for availability
    Log in for availability
    1 * 50 Feet
    1 * 50 Feet
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06509-13
    Model
    06509-13
    Length per pack
    15,2 m (50 ft.)
    Max. pressure
    2,7 bar (40 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8,8 m (29 ft.)
    Warranty
    1 year
    CE certificate
  • Tygon E-Lab (E-3603) Pump Tubing, L/S 14, 50 ft.
    1,60 mm (0,063")
    Tygon® E-Lab (E-3603)
    14
    0,0625"
    130 ml/min
    Log in for availability
    Log in for availability
    1 * 50 Feet
    1 * 50 Feet
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06509-14
    Model
    06509-14
    Length per pack
    15,2 m (50 ft.)
    Max. pressure
    2,7 bar (40 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8,8 m (29 ft.)
    Warranty
    1 year
    CE certificate
  • Tygon E-Lab (E-3603) Pump Tubing, L/S 16, 50 ft.
    3,10 mm (0,122")
    Tygon® E-Lab (E-3603)
    16
    0,125"
    480 ml/min
    Log in for availability
    Log in for availability
    1 * 50 Feet
    1 * 50 Feet
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06509-16
    Model
    06509-16
    Length per pack
    15,2 m (50 ft.)
    Max. pressure
    2,7 bar (40 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8,8 m (29 ft.)
    Warranty
    1 year
    CE certificate
  • Tygon E-Lab (E-3603) Pump Tubing, L/S 17, 50 ft.
    6,40 mm (0,252")
    Tygon® E-Lab (E-3603)
    17
    0,25"
    1700 ml/min
    Log in for availability
    Log in for availability
    1 * 50 Feet
    1 * 50 Feet
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06509-17
    Model
    06509-17
    Length per pack
    15,2 m (50 ft.)
    Max. pressure
    1,4 bar (20 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    510 mm Hg (20" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    6,7 m (22 ft.)
    Warranty
    1 year
    CE certificate
  • Tygon E-Lab (E-3603) Pump Tubing, L/S 18, 50 ft.
    7,90 mm (0,311")
    Tygon® E-Lab (E-3603)
    18
    0,375"
    2300 ml/min
    Log in for availability
    Log in for availability
    1 * 50 Feet
    1 * 50 Feet
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06509-18
    Model
    06509-18
    Length per pack
    15,2 m (50 ft.)
    Max. pressure
    1 bar (15 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    510 mm Hg (20" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    6,7 m (22 ft.)
    Warranty
    1 year
    CE certificate
  • Tygon E-Lab (E-3603) Pump Tubing, L/S 25, 50 ft.
    4,80 mm (0,189")
    Tygon® E-Lab (E-3603)
    25
    0,1875"
    1000 ml/min
    Log in for availability
    Log in for availability
    1 * 50 Feet
    1 * 50 Feet
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06509-25
    Model
    06509-25
    Length per pack
    15,2 m (50 ft.)
    Max. pressure
    2,4 bar (35 psi)
    Min. temperature
    –46 °C (–51 °F)
    Max. temperature
    74 °C (165 °F)
    Min. temperature (dynamic)
    0 °C (32 °F)
    Max. temperature (dynamic)
    40 °C (104 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;EU Food 10/2011;FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Suction lift H₂O
    8,8 m (29 ft.)
    Warranty
    CE certificate