Masterflex® DB-9 Pump Interface Cables, Avantor®
Interface cable with DB9 male/DB9 female connectors
Catalog # MFLX22050-54
Supplier: Avantor Fluid Handling
Masterflex® DB-9 Pump Interface Cables, Avantor®
Catalog # MFLX22050-54
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Optimize performance of your Masterflex® pump.
- Suitable for use with Masterflex® L/S® drives with a 9-pin connector
Use these 9-pin interface cable assemblies with Masterflex L/S drives having a 9-pin connector; including the 07550- and 07551-series computer-compatible drives. Cable measures 6 ft (1.8 m) long.
Specifications
- 説明:Interface cable with DB9 male/DB9 female connectors
- Cat. No.:MFLX22050-54
- Warranty:LTD life time
- Model:22050-54