Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
About this item
Create your own flowmeter systems for complete flexibility.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
- Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
- Use with media for laboratory and industrial applications
- Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
- Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
- Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.
37 Options Available Below
Product Details & Documents
Create your own flowmeter systems for complete flexibility.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
- Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
- Use with media for laboratory and industrial applications
- Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
- Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
- Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX03214-10
- MFLX03214-12
- MFLX03214-16
- MFLX03214-18
- MFLX03214-28
- MFLX03214-30
- MFLX03214-36
- MFLX03214-46
- MFLX03214-50
- MFLX03214-52
- MFLX03214-56
- MFLX03214-62
- MFLX03214-64
- MFLX03214-66
- MFLX03214-68
- MFLX03214-74
- MFLX03214-76
- MFLX03214-78
- MFLX03214-80
- MFLX03215-10
- MFLX03215-12
- MFLX03215-16
- MFLX03215-18
- MFLX03215-28
- MFLX03215-30
- MFLX03215-34
- MFLX03215-36
- MFLX03215-46
- MFLX03215-50
- MFLX03215-52
- MFLX03215-56
- MFLX03215-64
- MFLX03215-68
- MFLX03215-74
- MFLX03215-76
- MFLX03215-78
- MFLX03215-80
- Max. operating temperature:
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifications
Product Family Options
Product Information
- 説明Process connectionAvailabilityPrice
- Two Tube Frame for 65 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-10Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-12Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-16Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-18Max. operating temperature121 °C (250 °F)Warranty1 year - Three Tube Frame for 65 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-28Max. operating temperature121 °C (250 °F)Warranty1 year - Three Tube Frame for 65 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-30Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-36Max. operating temperature121 °C (250 °F)Warranty1 year - Four Tube Frame for 65 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-46Max. operating temperature121 °C (250 °F)Warranty1 year - Four Tube Frame for 65 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-50Max. operating temperature121 °C (250 °F)Warranty1 year - Four Tube Frame for 65 mm Flowtube, Isolated, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-52Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-56Max. operating temperature121 °C (250 °F)Warranty1 year - Five Tube Frame for 65 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-62Max. operating temperature121 °C (250 °F)Warranty1 year - Five Tube Frame for 65 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-64Max. operating temperature121 °C (250 °F)Warranty1 year - Five Tube Frame for 65 mm Flowtube, Isolated, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-66Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-68Max. operating temperature121 °C (250 °F)Warranty1 year - Six Tube Frame for 65 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03214-74Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-76Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-78Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03214-80Max. operating temperature121 °C (250 °F)Warranty1 year - Two Tube Frame for 150 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-10Max. operating temperature121 °C (250 °F)Warranty1 year - Two Tube Frame for 150 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-12Max. operating temperature121 °C (250 °F)Warranty1 year - Two Tube Frame for 150 mm Flowtube, Isolated, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-16Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03215-18Max. operating temperature121 °C (250 °F)Warranty1 year - Three Tube Frame for 150 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-28Max. operating temperature121 °C (250 °F)Warranty1 year - Three Tube Frame for 150 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-30Max. operating temperature121 °C (250 °F)Warranty1 year - Three Tube Frame for 150 mm Flowtube, Isolated, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-34Max. operating temperature121 °C (250 °F)Warranty1 year - Three Tube Frame for 150 mm Flowtube, Common, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-36Max. operating temperature121 °C (250 °F)Warranty1 year - Four Tube Frame for 150 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-46Max. operating temperature121 °C (250 °F)Warranty1 year - Four Tube Frame for 150 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-50Max. operating temperature121 °C (250 °F)Warranty1 year - Four Tube Frame for 150 mm Flowtube, Isolated, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-52Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03215-56Max. operating temperature121 °C (250 °F)Warranty1 year - Five Tube Frame for 150 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-64Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03215-68Max. operating temperature121 °C (250 °F)Warranty1 year - Six Tube Frame for 150 mm Flowtube, Isolated, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-74Max. operating temperature121 °C (250 °F)Warranty1 year - Six Tube Frame for 150 mm Flowtube, Common, Aluminum0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-76Max. operating temperature121 °C (250 °F)Warranty1 year - Six Tube Frame for 150 mm Flowtube, Isolated, 316 SS0,125" NPT(F)Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX03215-78Max. operating temperature121 °C (250 °F)Warranty1 year - Specifications:
More
Specifications
Cat. No.MFLX03215-80Max. operating temperature121 °C (250 °F)Warranty1 year