Masterflex® Self-Priming Micro Pumps, Avantor®
About this item
Accurate volumetric dispensing of 4 microliters to 250 microliters.
- Positive shut with pressures less than 3 psi
- High cycle life - up to 20 million dispense cycles
- Low power consumption
These pumps are factory set for discrete outputs ranging from 4ìl to 250ìl. The pumps can be cycled at up to 2.0 Hz for the 73120-1* and -2* series of pumps. All the while the pump can maintain a high level of accuracy with a deviation from set-point being less than 1%.
The pumps are self-priming. Intially, the suction created by the larger pumps is sufficient to pull liquids from an unpressurized container located up to four feet below the. Once primed, the pump is able to maintain around 5 psi pressure.
Long life (3,000 hours) is built into each pump with a guarantee for up to 20 million actuations.
14 Options Available Below
Product Details & Documents
Accurate volumetric dispensing of 4 microliters to 250 microliters.
- Positive shut with pressures less than 3 psi
- High cycle life - up to 20 million dispense cycles
- Low power consumption
These pumps are factory set for discrete outputs ranging from 4ìl to 250ìl. The pumps can be cycled at up to 2.0 Hz for the 73120-1* and -2* series of pumps. All the while the pump can maintain a high level of accuracy with a deviation from set-point being less than 1%.
The pumps are self-priming. Intially, the suction created by the larger pumps is sufficient to pull liquids from an unpressurized container located up to four feet below the. Once primed, the pump is able to maintain around 5 psi pressure.
Long life (3,000 hours) is built into each pump with a guarantee for up to 20 million actuations.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX73120-10
- MFLX73120-12
- MFLX73120-14
- MFLX73120-16
- MFLX73120-20
- MFLX73120-22
- MFLX73120-24
- MFLX73120-30
- MFLX73120-32
- MFLX73120-34
- MFLX73120-36
- MFLX73120-38
- MFLX73120-40
- MFLX73120-64
- Model:
- 73120-10
- 73120-12
- 73120-14
- 73120-16
- 73120-20
- 73120-22
- 73120-24
- 73120-30
- 73120-32
- 73120-34
- 73120-36
- 73120-38
- 73120-40
- 73120-64
- Inlet connection:
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- Max. temperature:
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- Outlet connection:
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- 1/4"-28 UNF(F)
- Power pigtail connection:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Run dry:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Self-priming:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Suction lift, wet:
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- 1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)
- Wetted materials:
- PPS, EPDM
- PPS, EPDM
- PPS, EPDM
- PPS, EPDM
- PPS, EPDM
- PPS, EPDM
- PPS, EPDM
- PTFE, Perfluorelastomer
- PTFE, Perfluorelastomer
- PTFE, Perfluorelastomer
- PTFE, Perfluorelastomer
- PTFE, Perfluorelastomer
- PTFE, Perfluorelastomer
- PTFE, Perfluorelastomer
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Specifications
Product Family Options
Product Information
- 説明最大流量電力高さ長さ幅AvailabilityPrice
- Self-Priming Micro Pump; 20 uL, PPS/EPDM, 12 VDC2,4 ml/min12 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-10Model73120-10Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 20 uL, PPS/EPDM, 24 VDC2,4 ml/min24 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-12Model73120-12Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 30 uL, PPS/EPDM, 12 VDC3,6 ml/min12 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-14Model73120-14Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 30 uL, PPS/EPDM, 24 VDC3,6 ml/min24 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-16Model73120-16Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 40 uL, PPS/EPDM, 24 VDC4,8 ml/min24 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-20Model73120-20Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 50 uL, PPS/EPDM, 12 VDC6 ml/min12 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-22Model73120-22Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 50 uL, PPS/EPDM, 24 VDC6 ml/min24 VDC2,54 cm6,35 cm3,65 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-24Model73120-24Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPPS, EPDMWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 20 uL, PTFE/PTFE, 12 VDC2,4 ml/min12 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-30Model73120-30Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 20 uL, PTFE/PTFE, 24 VDC2,4 ml/min24 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-32Model73120-32Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 30 uL, PTFE/PTFE, 12 VDC3,6 ml/min12 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-34Model73120-34Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 30 uL, PTFE/PTFE, 24 VDC3,6 ml/min24 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-36Model73120-36Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 40 uL, PTFE/PTFE, 12 VDC4,8 ml/min12 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-38Model73120-38Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 40 uL, PTFE/PTFE, 24 VDC4,8 ml/min24 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:CE Compliant
More
Specifications
Cat. No.MFLX73120-40Model73120-40Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 yearsCE Compliant - Self-Priming Micro Pump; 250 uL, PPS/EPDM, 24 VDC30 ml/min24 VDC2,54 cm7,14 cm3,81 cmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.MFLX73120-64Model73120-64Inlet connection1/4"-28 UNF(F)Max. temperature70 °C (158 °F)Outlet connection1/4"-28 UNF(F)Power pigtail connectionYesRun dryYesSelf-primingYesSuction lift, wet1200 mm of H₂O (48" H₂O); 671 mm Hg (3,5" Hg)Wetted materialsPTFE, PerfluorelastomerWarranty2 years