Stopcocks with Flow Indicator, PVC
Supplier: Avantor Fluid Handling
About this item
Easily control flow with flow indicator handle.
- Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.
4 Options Available Below
Product Details & Documents
Easily control flow with flow indicator handle.
- Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.
Specifications for Product Family
Specifications
- Connection type:
- NPT(M)
- NPT(M) x Barbed
- Barbed
- NPT(F)
- Inlet port:
- 1/4" NPT(M)
- 1/4" NPT(M)
- 1/4" to 1/2" Barb
- 1/4" NPT(F)
- Outlet port:
- 1/4" NPT(M)
- 1/4" Barb
- 1/4" to 1/2" Barb
- 1/4" NPT(F)
- Int. Ø tubing:
- 0,5 - 0,625"
- 0,25 - 0,5"
- Max. pressure:
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- Max. temperature:
- 48 °C (120 °F)
- 48 °C (120 °F)
- 48 °C (120 °F)
- 48 °C (120 °F)
- Port size:
- 0,25 NPT(F)
- Orifice:
- 6,35 mm (0,25")
- 6,35 mm (0,25")
- 6,35 to 12,7 mm (0,25 - 0,5")
- 6,35 mm (0,25")
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- Wetted materials:
- PVC
- PVC
- PVC
- PVC
Specifications
Product Family Options
Product Information
- 説明AvailabilityPrice
- Specifications:
More
Specifications
Cat. No.MFLX06225-52Connection typeNPT(M)Inlet port1/4" NPT(M)Outlet port1/4" NPT(M)Int. Ø tubingMax. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6,35 mm (0,25")Warranty1 yearWetted materialsPVC - Specifications:
More
Specifications
Cat. No.MFLX06225-54Connection typeNPT(M) x BarbedInlet port1/4" NPT(M)Outlet port1/4" BarbInt. Ø tubing0,5 - 0,625"Max. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6,35 mm (0,25")Warranty1 yearWetted materialsPVC - Specifications:
More
Specifications
Cat. No.MFLX06225-56Connection typeBarbedInlet port1/4" to 1/2" BarbOutlet port1/4" to 1/2" BarbInt. Ø tubing0,25 - 0,5"Max. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6,35 to 12,7 mm (0,25 - 0,5")Warranty1 yearWetted materialsPVC - Specifications:
More
Specifications
Cat. No.MFLX06225-50Connection typeNPT(F)Inlet port1/4" NPT(F)Outlet port1/4" NPT(F)Int. Ø tubingMax. pressure150 psiMax. temperature48 °C (120 °F)Port size0,25 NPT(F)Orifice6,35 mm (0,25")Warranty1 yearWetted materialsPVC