Masterflex® Ismatec® Quick-Couple Pump Head for 1.6 mm Wall Tubing, 3 Rollers, 3400 mL/min
Catalog # MFLX78002-16
Supplier: Avantor Fluid Handling
Specifications
- 最大流量:3400 ml/min
- Max. pressure:1,5 bar (21,7 psi)
- Min. flow rate:0,45 ml/min
- No. of channels:1
- 説明:Quick-Couple pump head with 3 rollers, 1,6 mm wall tubing, 3400 ml/min max flow
- Seq:04
- Model:78002-16
- Warranty:2 years
- Series:Quick-Coupled
- Pk:1
- Rollers:3
Specifications
About this item
Snap your pump head onto your drives quickly with turn-and-lock mounting system.
- Select from interchangeable pump heads that offer flow rates from 0.01 to 3660 ml/min
- 3 rollers
Ensure optimum performance from your Ismatec® pumps.
- 最大流量:3400 ml/min
- Max. pressure:1,5 bar (21,7 psi)
- Min. flow rate:0,45 ml/min
- No. of channels:1
- 説明:Quick-Couple pump head with 3 rollers, 1,6 mm wall tubing, 3400 ml/min max flow
- Seq:04
- Environmentally Preferable:
- Model:78002-16
- Warranty:2 years
- Series:Quick-Coupled
- Pk:1
- Rollers:3