Flux® ドラムポンプチューブセット、PVDF
Supplier: Avantor Fluid Handling
Certificates
About this item
Construct a pump system that meets your specific needs.
- PVDF sets are ideal for your high-purity applications
Standard, High-Head, and Heavy-Duty Tube Sets feature a sealless design for low maintenance and long life. All tube sets include a hose barb fitting made of the same material as the tube set. Fitting accepts either 3/4" or 1" ID tubing.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX70601-04
- MFLX70601-00
- MFLX70601-02
- Max. temperature:
- 212 °C
- 212 °C
- 212 °C
- Outlet connection:
- 3/4" hose barb
- 3/4" hose barb
- 3/4" hose barb
- Tube ext.Ø, inlet:
- 4 cm (1,5625")
- 4 cm (1,5625")
- 4 cm (1,5625")
- Tube length, inlet:
- 47"
- 27"
- 39"
- Wetted materials:
- PVDF body; Tefzel impeller, Viton, PTFE, carbon, Hastelloy C
- PVDF body; Tefzel impeller, Viton, PTFE, carbon, Hastelloy C
- PVDF body; Tefzel impeller, Viton, PTFE, carbon, Hastelloy C
- Warranty:
- 6 Months
- 6 Months
- 6 Months