Orders LinkContactUsLinkComponent
Masterflex® Flow Sensor Flow Controllers, Avantor®
Masterflex® Flow Sensor Flow Controllers, Avantor®
Masterflex® Flow Sensor Flow Controllers, Avantor®
Masterflex® Flow Sensor Flow Controllers, Avantor®
Easily control, view, and store your flow rates.
  • Control your Low-Flow Impeller Sensors with these controllers
These new controllers display flow rate and total (resettable) simultaneously on two 8-digit Display LCDs. Push-button keys and menu prompts make it easy to program the controller. The 4-to-20 mA output lets you send data to a recorder or datalogger. Nonvolatile memory stores flow total in the event of power failure.
Select a 1¼4 DIN panel- or wall-mount controller depending on your application needs. Panel-mount model 33110-60 features a gasketed front panel, membrane switches, and NEMA Type 12 (IP52) enclosure. Wall-mount model 33110-70 has a clear cover and splashproof NEMA Type 4X (IP56) enclosure. Both models include a 21¼2-ft L cable with stripped ends. Order power supply for 110 VAC operation separately.
  • Display type:
  • Rate 5 digit autorange, Total 8 digit
  • Rate 5 digit autorange, Total 8 digit
  • Enclosure rating:
  • NEMA 4X, IP67
  • NEMA 4X, IP67
  • Input:
  • 5V pulse or contact closure
  • 5V pulse or contact closure
  • Max. operating temperature:
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • Accuracy:
  • ±0,2% of reading
  • ±0,2% of reading
  • Warranty:
  • 2 years
  • 2 years
Product Family Options
  • 説明
    出力
    電力
  • Panel Mount Flow Rate/Total Indicator, Pulse Input, 4-20 mA Output
    12 to 24 VDC; K-factor: 0,050 to 1999,999
    7 - 30 VDC, 4 mA (4-20 mA whenloop-powered)
    1 * 1 items
    1 * 1 items
    2 year warranty
    Specifications
    Cat. No.
    MFLX33110-60
    Display type
    Rate 5 digit autorange, Total 8 digit
    Enclosure rating
    NEMA 4X, IP67
    Input
    5V pulse or contact closure
    Max. operating temperature
    70 °C (158 °F)
    Accuracy
    ±0,2% of reading
    Warranty
    2 years
    2 year warranty
  • Wall Mount Flow Rate/Total Indicator, Pulse Input, 4-20 mA Output
    12 to 24 VDC; K-factor: 0,050 to 1999,999
    7 - 30 VDC, 4 mA (4-20 mA whenloop-powered)
    1 * 1 items
    1 * 1 items
    2 year warranty
    Specifications
    Cat. No.
    MFLX33110-70
    Display type
    Rate 5 digit autorange, Total 8 digit
    Enclosure rating
    NEMA 4X, IP67
    Input
    5V pulse or contact closure
    Max. operating temperature
    70 °C (158 °F)
    Accuracy
    ±0,2% of reading
    Warranty