Blue-White® Direct-Read Variable-Area Flowmeters for Water, Panel-Mount
Get better results than acrylic.
- Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get a flow rate
- Use with water for laboratory and industrial applications
These panel-mount flowmeters feature rotatable 90° elbow adapters for panel-mount installation. Installation is easy—simply drill two holes in your panel, insert meter fittings through the holes, secure with lock nuts (included), and rotate scale for easy viewing.
Get better results than acrylic.
- Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get a flow rate
- Use with water for laboratory and industrial applications
These panel-mount flowmeters feature rotatable 90° elbow adapters for panel-mount installation. Installation is easy—simply drill two holes in your panel, insert meter fittings through the holes, secure with lock nuts (included), and rotate scale for easy viewing.
- Model:
- 32471-00
- 32471-01
- 32471-02
- 32471-03
- 32471-04
- Accuracy:
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- Float material:
- PVC
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- Max. operating temperature:
- 115,5 °C (240 °F)
- 115,5 °C (240 °F)
- 115,5 °C (240 °F)
- 115,5 °C (240 °F)
- 115,5 °C (240 °F)
- Max. pressure:
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Product Family Options
- 説明最小流量最大流量Media typeValveHousing materialProcess connection
- Variable-Area Flowmeter, Panel-Mount, Direct Reading, Polysulfone Housing, 0,25 gal./min Water0,1 L/min (0,025 gal./min)1 L/min (0,25 gal./min)WaterNoPSF0,5" NPT(M)1 * 1 items
Specifications
Cat. No.MFLX32471-00Model32471-00Accuracy±5% full-scaleFloat materialPVCMax. operating temperature115,5 °C (240 °F)Max. pressure150 psiWarranty1 year - Variable-Area Flowmeter, Panel-Mount, Direct Reading, Polysulfone Housing, 1 gal./min Water0,4 L/min (0,1 gal./min)4 L/min (1 gal./min)WaterNoPSF0,5" NPT(M)1 * 1 items
Specifications
Cat. No.MFLX32471-01Model32471-01Accuracy±5% full-scaleFloat material316 Stainless SteelMax. operating temperature115,5 °C (240 °F)Max. pressure150 psiWarranty1 year - Variable-Area Flowmeter, Panel-Mount, Direct Reading, Polysulfone Housing, 2 gal./min Water0,75 L/min (0,2 gal./min)7,5 L/min (2 gal./min)WaterNoPSF0,5" NPT(M)1 * 1 items
Specifications
Cat. No.MFLX32471-02Model32471-02Accuracy±5% full-scaleFloat material316 Stainless SteelMax. operating temperature115,5 °C (240 °F)Max. pressure150 psiWarranty1 year - Variable-Area Flowmeter, Panel-Mount, Direct Reading, Polysulfone Housing, 5 gal./min Water1,8 L/min (0,5 gal./min)18 L/min (5 gal./min)WaterNoPSF0,5" NPT(M)1 * 1 items
Specifications
Cat. No.MFLX32471-03Model32471-03Accuracy±5% full-scaleFloat material316 Stainless SteelMax. operating temperature115,5 °C (240 °F)Max. pressure150 psiWarranty1 year - Variable-Area Flowmeter, Panel-Mount, Direct Reading, Polysulfone Housing, 10 gal./min Water5 L/min (1 gal./min)37,5 L/min (10 gal./min)WaterNoPSF0,75" NPT(M)1 * 1 items
Specifications
Cat. No.MFLX32471-04Model32471-04Accuracy±5% full-scaleFloat material316 Stainless SteelMax. operating temperature115,5 °C (240 °F)Max. pressure150 psiWarranty1 year