Masterflex® Dispensing Tip with Luer Lock, Polypropylene, Luer Lock by 1/16" to 1/8" ID
MFLX78226-62
- 接続タイプ:Female Luer to Barb
- Flow pattern:Straight
- 説明:Polypropylene dispensing tip with Luer lock for ¹⁄₁₆" and ⅛" ID tubing.
- Seq:01
- Pk:1
- Fitting material:Polypropylene
Attach with just a twist!
- Constructed of durable polypropylene
- Easily attach flexible tubing to the hose barb connection
These luer fittings are ideal for a variety of critical applications, including connection to syringes or small diameter flexible tubing.
- 接続タイプ:Female Luer to Barb
- Flow pattern:Straight
- 説明:Polypropylene dispensing tip with Luer lock for ¹⁄₁₆" and ⅛" ID tubing.
- Seq:01
- Environmentally Preferable:
- Pk:1
- Fitting material:Polypropylene