Masterflex® Connection cable, "D" connector to stripped ends, 3-ft
Catalog # MFLX98651-50
Supplier: Avantor Fluid Handling
Specifications
- 説明:Connection cable, "D" connector to stripped ends
- 長さ:3 ft
- Model:98651-50
- Warranty:1 year
- Pk:3 Feet
Specifications
About this item
Easy connect to your solenoid valve.
- Optimize performance of your Masterflex® Solenoid Valves
"D" connector connection cable with stripped wire ends for Aalborg stepping motor proportioning valves. This cable is 3-ft (0.9m) long.
- 説明:Connection cable, "D" connector to stripped ends
- 長さ:3 ft
- Environmentally Preferable:
- Model:98651-50
- Warranty:1 year
- Pk:3 Feet