Orders LinkContactUsLinkComponent
VWR®, プラットフォームスケール
VWR®, プラットフォームスケール
Supplier:   VWR Collection
VWR®, プラットフォームスケール
Supplier:  VWR Collection
Certificates
VWR®, プラットフォームスケール
VWR®, プラットフォームスケール
12 Options Available Below
Product Details & Documents
Industrial series of advanced platform scales, offering increased accuracy and fast stabilisation time, combined with easy, intuitive operation and a large graphical screen for laboratory applications and industrial settings. Ideal for weighing, mixing, counting and check weighing.
  • Efficient filtering algorithms to deliver quick and accurate results
  • Fast stabilisation time
  • Easy to clean and high chemical resistance
  • Choice of two platform sizes (M-Line 280×280 mm, L-Line 300×400 mm)
  • Different column size, for tabletop or ground weighing (M-Line 360 mm, L-Line 660 mm)
  • Nine built-in applications with GLP/GMP compliant data output: Weighing and dosing - counting - percentage weighing - density determination - measuring units - animal weighing - formulation - totalisation
  • GLP/ISO record keeping of weight values with date, time and serial number
  • Connectivity RS232 - printer and PC
Certifications: IP Protection Class according to DIN EN 60529: IP 54
納品情報: Supplied with main adapter and universal plug. Calibration weights are available from Avantor. Please see 'Calibration weights'.
Product Family Options
M series models
Product Information
  • モデル
    Weighing cap.
    Readability
    パン/プラットフォームサイズ
    Repeatability
    Linearity
    Availability
    Price
  • IS-PMG 3201
    3200 g
    0,1 g
    280×280 mm
    0,05 g
    ±0,2 g
    1 * 1 items
  • IS-PMG 5201
    5200 g
    0,1 g
    280×280 mm
    0,05 g
    ±0,2 g
    1 * 1 items
  • IS-PMG 6200
    6200 g
    1 g
    280×280 mm
    0,1 g
    ±0,1 g
    1 * 1 items
  • IS-PMG 10001
    10000 g
    0,1 g
    280×280 mm
    0,05 g
    ±0,2 g
    1 * 1 items
  • IS-PMG 10000
    10000 g
    1 g
    280×280 mm
    0,1 g
    ±0,1 g
    1 * 1 items
  • IS-PMG 15001
    15000 g
    0,1 g
    280×280 mm
    0,08 g
    ±0,3 g
    1 * 1 items
  • IS-PMG 20001
    20000 g
    0,1 g
    280×280 mm
    0,1 g
    ±0,3 g
    1 * 1 items
  • IS-PMG 20000
    20000 g
    1 g
    280×280 mm
    0,1 g
    ±0,1 g
    1 * 1 items
L series models
Product Information
  • モデル
    Weighing cap.
    Readability
    パン/プラットフォームサイズ
    Repeatability
    Linearity
    Availability
    Price
  • IS-PLG 35001
    35000 g
    0,1 g
    300×400 mm
    0,1 g
    ±0,4 g
    1 * 1 items
  • IS-PLG 10000
    10000 g
    1 g
    300×400 mm
    0,1 g
    ±0,1 g
    1 * 1 items
  • IS-PLG 20000
    20000 g
    1 g
    300×400 mm
    0,1 g
    ±0,1 g
    1 * 1 items
  • IS-PLG 60000
    60000 g
    1 g
    300×400 mm
    0,1 g
    ±0,1 g
    1 * 1 items
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 611-4790
    TX-110 dot printer for balances
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 611-2786
    Serial RS232 to USB converter
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 611-3824
    Complete communication kit with i-Weight software to connect a balance to a computer. Enables data collection, statistics, graphs and exports results to Excel. Functions: Piece counting, Density, Peak hold, Limits set. OS: Windows 7, 8, 10 (32&64). Note not suitable for use with moisture analysers
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 611-2787
    Serial cable 9 pins M/M for serial output to printer