Orders LinkContactUsLinkComponent
酸化還元(ORP)標準
酸化還元(ORP)標準
酸化還元(ORP)標準
Supplier:  VWR Chemicals
Certificates
酸化還元(ORP)標準
酸化還元(ORP)標準
13 Options Available Below
Product Details & Documents
Avantor offers a wide range of REDOX (ORP) standards, which are produced from high quality reagents.These reference materials can be used as control standards to check the functionality of both the elextrodes and the measuring instruments. The values are temperature dependent, so it is important to control the environment of the measurement. A table of temperatue and corresponding Redox value is available on both the label and the batch specific certificate.
  • Accuracy ±5 mV
  • Quoted values tested at 25 °C
  • Temperature vs Redox value as a table on the bottle label
  • Stability excellent throughout shelf life
ORP standards where all values quoted are potentials of platinum electrode v Ag/AgCl reference electrode (3M KCl)
Documents
Product Family Options
Product Information
  • 説明
    Accuracy
    パックタイプ
    Pk
    Availability
    Price
  • 124 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89
  • 200 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89
  • 220 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.80
    $205.80
    1 * 500 mL
       $205.80
  • 250 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89
  • 300 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89
  • 358 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $216.21
    $216.21
    1 * 500 mL
       $216.21
  • 400 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $216.21
    $216.21
    1 * 500 mL
       $216.21
  • 440 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $216.21
    $216.21
    1 * 500 mL
       $216.21
  • 465 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.54
    $205.54
    1 * 500 mL
       $205.54
  • 468 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.00
    $205.00
    1 * 500 mL
       $205.00
  • 475 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89
  • 600 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89
  • 650 mV (at 25°C)
    ±5 mV
    HDPE bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - $205.89
    $205.89
    1 * 500 mL
       $205.89