Orders LinkContactUsLinkComponent
VWR®, シンチレーションバイアル
VWR®, シンチレーションバイアル
VWR®, シンチレーションバイアル
Supplier:  VWR Collection
Certificates
VWR®, シンチレーションバイアル
VWR®, シンチレーションバイアル
2 Options Available Below
Product Details & Documents
VWR® scintillation vials are produced from borosilicate 5.1 expansion glass. They are compatible with all scintillation counters.
  • High resistance against chemicals and thermal shock
  • Low background counts and high ultraviolet transmission
  • Delivered in convenient pack sizes
VWR® scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting or for sample storage.
Ordering information: Screw caps have to be ordered separately and you can choose the right screw cap for your application from our selection of fitting screw caps with the most common liners, which we offer as accessories.
Packaging: Packed in shrink-wrapped trays in a convenient 250 piece pack size.
Product Family Options
Vials
Product Information
  • 容量
    Ø×H
    Colour
    キャップ付き
    キャップサイズ
    Availability
    Price
  • 6,00 ml
    17×58 mm
    Clear
    No
    15-425
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • 20,00 ml
    28×61 mm
    Clear
    No
    22-400
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
Screw caps
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, UREA, with metal foil/pulp liner
    15-425
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, PP, with metal foil/pulp liner
    22-400
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, UREA, with metal foil/pulp liner
    22-400
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, UREA, with PE cone liner
    22-400
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, PP, with metal foil/pulp liner
    15-425
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, PP, with foamed PE liner
    22-400
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Product Information
  • Catalog #
    Type
    キャップサイズ
    Availability
    Price
  • Screw cap, UREA, with PE disc liner
    22-400
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00