Orders LinkContactUsLinkComponent
マイクロチューブラック、J.T.Baker®、リバーシブルマイクロチューブラック
 :   Avantor
マイクロチューブラック、J.T.Baker®、リバーシブルマイクロチューブラック
 :  Avantor
マイクロチューブラック、J.T.Baker®、リバーシブルマイクロチューブラック
マイクロチューブラック、J.T.Baker®、リバーシブルマイクロチューブラック
Safe sample storage system.
  • V bottom for optimal sample recovery
  • Individual tubes or strips of eight tubes available
  • 96-well capmat and strips of eight plug caps available
  • Refillable alphanumeric marked rack
  • Non sterile
Microtube racks are supplied with tubes included. Rack can be also filled with individual tubes available separately. They have a footprint in accordance to SBS standard and it is compatible with all current robotic systems.
  • Catalog No:
  • 45000-0000
  • 45005-0000
  • 45003-0000
  • 45000-0008
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Product Family Options
Microtube racks
  • 説明
    容量
    Colour
    Material
  • Microtube rack with 96 individual tubes
    1,2 ml
    Clear/blue
    PP
    1 * 10 items
    1 * 10 items - $241.84
    $241.84
    1 * 10 items
       $241.84
      Low Manufacturing Impact
    Specifications
    Cat. No.
    45000-0000
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • Microtube rack with strips of 8 tubes
    1,2 ml
    Clear/blue
    PP
    1 * 10 items
    1 * 10 items - $247.18
    $247.18
    1 * 10 items
       $247.18
      Low Manufacturing Impact
    Specifications
    Cat. No.
    45005-0000
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Tubes only
  • 説明
    容量
    Colour
    Material
  • Individual tubes
    1,2 ml
    Clear
    PP
    1 * 960 items
    1 * 960 items - $154.78
    $154.78
    1 * 960 items
       $154.78
    Specifications
    Cat. No.
    45003-0000
    Environmentally Preferable
    Environmental certification
  • Strip of 8 tubes
    1,2 ml
    Clear
    PP
    1 * 120 items
    1 * 120 items - $128.95
    $128.95
    1 * 120 items
       $128.95
      Low Manufacturing Impact
    Specifications
    Cat. No.
    45000-0008
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Caps only
  •  
    Colour
    説明
  • Clear
    Strip of 8 plug caps
    1 * 120 items
    1 * 120 items - $70.98
    $70.98
    1 * 120 items
       $70.98
Capmat
  •  
    Sterile
    Packed
    Material
    光学特性
    説明
  • -
    1 bag of 20
    EVA
    Transparent
    Capmat for J.T.Baker Plate Round Well
    1 * 20 items
    1 * 20 items - $116.06
    $116.06
    1 * 20 items
       $116.06