VWR® 211DS, パーソナル型加振インキュベーター
Universal spring-loaded platform for flasks, bottles or tube arcks
Catalog # VWRIS2031-13
Supplier: VWR Collection
Jump to:
Product Details & Documents
A full range of accessories is available to complement the 211DS. The 211DS includes three shelves (one is a half-size shelf, allowing tall vessels to be accommodated). Two shaking platforms (sold separately) are available for the 211DS; the flask platform is predrilled to accept a variety of different sized clamps while the flat platform has a nonslip rubber mat for holding dishes and plates in place.
Specifications
- 説明:Universal spring-loaded platform for flasks, bottles or tube arcks
- Cat. No.:VWRIS2031-13