Orders LinkContactUsLinkComponent
VWR®ピペットチップ
VWR®ピペットチップ
Supplier:  VWR Collection
Certificates
VWR®ピペットチップ
VWR®ピペットチップ
47 Options Available Below
Product Details & Documents
These VWR® pipette tips are designed for precise liquid transfer.
  • Fully autoclavable to 121 °C
  • Colour-coded racks for easy volume identification
  • Tips with PP filter are also available
  • DNase-/RNase-free and non-pyrogenic
  • Every rack or case is printed with a batch number for traceability
  • Sterile tips are sterilized by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf Life: 3 years after month of production
Good quality consumable products for micropipettors. Preferred accessories for most brand micropipettors.
 : Manufactured in a class 100 000 room environment under ISO 13485 and ISO 9001 quality management system.
Product Family Options
VWR® pipette tips, natural
Product Information
  • Tip style
    容量
    Sterile
    Filter
    Colour code
    Packed
    Availability
    Price
  • Standard
    10 µl
    Grey/red
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $83.00
    $83.00
    1 * 10.000 items
       $83.00
  • Standard
    10 µl
    +
    Grey/red
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $185.36
    $185.36
    1 * 10.000 items
       $185.36
  • Extended length
    10 µl
    -
    -
    Neutral
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $87.71
    $87.71
    1 * 10.000 items
       $87.71
  • Standard
    10 µl
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $55.97
    $55.97
    1 * 1.920 items
       $55.97
  • Standard
    10 µl
    +
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $76.67
    $76.67
    1 * 1.920 items
       $76.67
  • Extended length
    10 µl
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $61.60
    $61.60
    1 * 1.920 items
       $61.60
  • Extended length
    10 µl
    +
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $84.33
    $84.33
    1 * 1.920 items
       $84.33
  • Standard
    20 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $338.71
    $338.71
    1 * 10.000 items
       $338.71
  • Standard
    20 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $91.59
    $91.59
    1 * 1.920 items
       $91.59
  • Standard
    100 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $338.71
    $338.71
    1 * 10.000 items
       $338.71
  • Standard
    100 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $91.57
    $91.57
    1 * 1.920 items
       $91.57
  • Standard
    200 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $104.49
    $104.49
    1 * 10.000 items
       $104.49
  • Standard
    200 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $252.77
    $252.77
    1 * 10.000 items
       $252.77
  • Standard
    200 µl
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $75.04
    $75.04
    1 * 1.920 items
       $75.04
  • Standard
    200 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $91.57
    $91.57
    1 * 1.920 items
       $91.57
  • Standard
    300 µl
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $106.15
    $106.15
    1 * 10.000 items
       $106.15
  • Standard
    300 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $252.77
    $252.77
    1 * 10.000 items
       $252.77
  • Standard
    300 µl
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $75.74
    $75.74
    1 * 1.920 items
       $75.74
  • Standard
    300 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $91.57
    $91.57
    1 * 1.920 items
       $91.57
  • Standard
    1000 µl
    Grey/blue
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $151.66
    $151.66
    1 * 10.000 items
       $151.66
  • Standard
    1000 µl
    +
    Grey/blue
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $303.29
    $303.29
    1 * 10.000 items
       $303.29
  • Standard
    1000 µl
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $97.30
    $97.30
    1 * 1.920 items
       $97.30
  • Standard
    1000 µl
    +
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $126.80
    $126.80
    1 * 1.920 items
       $126.80
  • Extended length
    1000 µl
    Grey/blue
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $187.46
    $187.46
    1 * 10.000 items
       $187.46
  • Extended length
    1000 µl
    +
    Grey/blue
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $374.93
    $374.93
    1 * 10.000 items
       $374.93
  • Extended length
    1000 µl
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $117.25
    $117.25
    1 * 1.920 items
       $117.25
  • Extended length
    1000 µl
    +
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $154.73
    $154.73
    1 * 1.920 items
       $154.73
  • Extended length
    1000 µl
    +
    -
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $138.51
    $138.51
    1 * 1.920 items
       $138.51
VWR® pipette tips, super low retention, natural
Product Information
  • Tip style
    容量
    Sterile
    Filter
    Colour code
    Packed
    Availability
    Price
  • Standard
    10 µl
    Grey/red
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $144.19
    $144.19
    1 * 10.000 items
       $144.19
  • Standard
    10 µl
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $64.35
    $64.35
    1 * 1.920 items
       $64.35
  • Standard
    10 µl
    +
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $89.13
    $89.13
    1 * 1.920 items
       $89.13
  • Extended length
    10 µl
    Grey/red
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $219.87
    $219.87
    1 * 10.000 items
       $219.87
  • Extended length
    10 µl
    +
    -
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $68.16
    $68.16
    1 * 1.920 items
       $68.16
  • Extended length
    10 µl
    +
    +
    Grey/red
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $90.66
    $90.66
    1 * 1.920 items
       $90.66
  • Standard
    20 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $403.68
    $403.68
    1 * 10.000 items
       $403.68
  • Standard
    20 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $105.53
    $105.53
    1 * 1.920 items
       $105.53
  • Standard
    100 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $439.74
    $439.74
    1 * 10.000 items
       $439.74
  • Standard
    100 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $105.53
    $105.53
    1 * 1.920 items
       $105.53
  • Standard
    200 µl
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $139.25
    $139.25
    1 * 10.000 items
       $139.25
  • Standard
    200 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $277.55
    $277.55
    1 * 10.000 items
       $277.55
  • Standard
    200 µl
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $89.50
    $89.50
    1 * 1.920 items
       $89.50
  • Standard
    200 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $105.55
    $105.55
    1 * 1.920 items
       $105.55
  • Standard
    300 µl
    +
    Grey/yellow
    1000 tips/bag, 10 bags/case
    Log in for availability
    Log in for availability
    1 * 10.000 items
    1 * 10.000 items - $277.55
    $277.55
    1 * 10.000 items
       $277.55
  • Standard
    300 µl
    +
    +
    Grey/yellow
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $105.55
    $105.55
    1 * 1.920 items
       $105.55
  • Standard
    1000 µl
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $113.49
    $113.49
    1 * 1.920 items
       $113.49
  • Standard
    1000 µl
    +
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $147.40
    $147.40
    1 * 1.920 items
       $147.40
  • Extended length
    1000 µl
    +
    +
    Grey/blue
    96 tips/rack, 10 racks/box, 2 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - $182.56
    $182.56
    1 * 1.920 items
       $182.56