VWR®, 半透明ユーロ容器
About this item
These sturdy, transparent plastic containers are made of translucent material. The translucent material allows users to check whether a container is empty or filled, even when stacked or closed.
- Made of translucent material PPC (polypropylene copolymer)
- Suitable for manual and automated handling of lightweight goods
- Smooth surfaces make them easy to clean
- Ergonomic handles for effective manual handling
- Suitable for –20 to +65 °C storage
- 100% recyclable
The translucent Euro containers are available in universal Euros sizes: 300×200, 400×300 and 600×400 mm. They are perfect for palletisation and very stable due to their double wall construction. They are suitable for every industry and for any application. They are also ideal for manual and automatic conveyor handling. The colour is natural translucent PP.
: Guaranteed food-safe.
: Trolleys and lids are optional and can be supplied for the entire range.
8 Options Available Below
Product Details & Documents
These sturdy, transparent plastic containers are made of translucent material. The translucent material allows users to check whether a container is empty or filled, even when stacked or closed.
- Made of translucent material PPC (polypropylene copolymer)
- Suitable for manual and automated handling of lightweight goods
- Smooth surfaces make them easy to clean
- Ergonomic handles for effective manual handling
- Suitable for –20 to +65 °C storage
- 100% recyclable
The translucent Euro containers are available in universal Euros sizes: 300×200, 400×300 and 600×400 mm. They are perfect for palletisation and very stable due to their double wall construction. They are suitable for every industry and for any application. They are also ideal for manual and automatic conveyor handling. The colour is natural translucent PP.
: Guaranteed food-safe.
: Trolleys and lids are optional and can be supplied for the entire range.
Product Family Options
Containers
Product Information
- 容量説明W×D×H ext.W×D×H int.AvailabilityPrice
- 2,9 LTranslucent Euro container, 15 kg load capacity, closed handles300×200×70 mm268×168×68 mmLog in for availability1 * 1 items
- 5,1 LTranslucent Euro container, 15 kg load capacity, closed handles300×200×120 mm268×168×118 mmLog in for availability1 * 1 items
- 7,2 LTranslucent Euro container, 15 kg load capacity, closed handles300×200×170 mm268×168×168 mmLog in for availability1 * 1 items
- 9,4 LTranslucent Euro container, 15 kg load capacity, closed handles300×200×220 mm268×168×218 mmLog in for availability1 * 1 items
- 20,8 LTranslucent Euro container, 25 kg load capacity, open handles400×300×220 mm369×270×218 mmLog in for availability1 * 1 items
- 23,9 LTranslucent Euro container, 25 kg load capacity, closed handles600×400×120 mm568×368×117 mmLog in for availability1 * 1 items
- 44,2 LTranslucent Euro container, 25 kg load capacity, open handles600×400×220 mm568×368×217 mmLog in for availability1 * 1 items
- 64,5 LTranslucent Euro container, 25 kg load capacity, open handles600×400×320 mm568×368×317 mmLog in for availability1 * 1 items
Lids
Product Information
- Catalog #説明寸法AvailabilityPrice