VWR®, パーフォレイテッド・ユーロコンテナー
About this item
These perforated plastic stackable Euro containers are made from heavy-duty polypropylene, suitable for every industry and for any application. They provide outstanding air and liquid permeability.
- Made of PPC (polypropylene copolymer)
- Optimal air circulation
- Hygienically flawless and easy to clean
- Ergonomic handles for effective manual handling
- Suitable for –20 to +65 °C storage
- 100% recyclable
The perforated Euro containers are available in universal Euros sizes: 400×300, 600×400 or 800×600 mm. They are perfect for palletisation. They are suitable for every industry and for any application. They are also suitable for manual and automatic conveyor handling. The standard colour is grey.
Certifications: Guaranteed food safe.
Ordering information: Trolleys and lids are optional and can be supplied for the entire range.
12 Options Available Below
Product Details & Documents
These perforated plastic stackable Euro containers are made from heavy-duty polypropylene, suitable for every industry and for any application. They provide outstanding air and liquid permeability.
- Made of PPC (polypropylene copolymer)
- Optimal air circulation
- Hygienically flawless and easy to clean
- Ergonomic handles for effective manual handling
- Suitable for –20 to +65 °C storage
- 100% recyclable
The perforated Euro containers are available in universal Euros sizes: 400×300, 600×400 or 800×600 mm. They are perfect for palletisation. They are suitable for every industry and for any application. They are also suitable for manual and automatic conveyor handling. The standard colour is grey.
Certifications: Guaranteed food safe.
Ordering information: Trolleys and lids are optional and can be supplied for the entire range.
Product Family Options
Containers
Product Information
- 容量説明W×D×H ext.W×D×H int.AvailabilityPrice
- 5,4 LPerforated Euro container, 25 kg load capacity, closed handles400×300×70 mm367×268×58 mmLog in for availability1 * 1 items
- 10,2 LPerforated Euro container, 25 kg load capacity, closed handles400×300×120 mm367×268×108 mmLog in for availability1 * 1 items
- 11,2 LPerforated Euro container, 25 kg load capacity, closed handles600×400×70 mm567×367×55 mmLog in for availability1 * 1 items
- 14,9 LPerforated Euro container, 25 kg load capacity, open handles400×300×170 mm367×268×158 mmLog in for availability1 * 1 items
- 19,8 LPerforated Euro container, 25 kg load capacity, open handles400×300×220 mm367×268×208 mmLog in for availability1 * 1 items
- 21,4 LPerforated Euro container, 25 kg load capacity, closed handles600×400×120 mm567×367×105 mmLog in for availability1 * 1 items
- 24,6 LPerforated Euro container, 25 kg load capacity, open handles400×300×270 mm367×268×258 mmLog in for availability1 * 1 items
- 29 LPerforated Euro container, 25 kg load capacity, open handles400×300×320 mm367×268×308 mmLog in for availability1 * 1 items
- 33,1 LPerforated Euro container, 25 kg load capacity, open handles600×400×170 mm567×367×155 mmLog in for availability1 * 1 items
- 41,7 LPerforated Euro container, 25 kg load capacity, open handles600×400×220 mm567×367205 mmLog in for availability1 * 1 items
- 51,9 LPerforated Euro container, 25 kg load capacity, open handles600×400×270 mm567×367×255 mmLog in for availability1 * 1 items
- 62 LPerforated Euro container, 25 kg load capacity, open handles600×400×320 mm567×367×305 mmLog in for availability1 * 1 items
Lids
Product Information
- Catalog #説明寸法AvailabilityPrice