VWR®, Solid Euro Containers
About this item
These plastic stackable Euro containers are ideal for simple storage solutions as well as highly automated systems. Made from strong plastic these containers are ideal for transporting goods and food safely. They are very stable when loaded due to reinforced frame and corner construction.
- Made of PPC (polypropylene copolymer)
- Suitable for every industry and for any application
- Smooth surfaces make them easy to clean
- Ergonomic handles for effective manual handling
- Suitable for –20 to +65 °C storage
- 100% recyclable
The solid Euro containers are available in universal Euros sizes: 300×200, 400×300, 600×400 or 800×600 mm. They are perfect for palletisation. They are also suitable for manual and automatic conveyor handling. The standard colour is grey.
Certifications: Guaranteed food-safe.
Ordering information: Trolleys and lids are optional and can be supplied for the entire range.
16 Options Available Below
Product Details & Documents
These plastic stackable Euro containers are ideal for simple storage solutions as well as highly automated systems. Made from strong plastic these containers are ideal for transporting goods and food safely. They are very stable when loaded due to reinforced frame and corner construction.
- Made of PPC (polypropylene copolymer)
- Suitable for every industry and for any application
- Smooth surfaces make them easy to clean
- Ergonomic handles for effective manual handling
- Suitable for –20 to +65 °C storage
- 100% recyclable
The solid Euro containers are available in universal Euros sizes: 300×200, 400×300, 600×400 or 800×600 mm. They are perfect for palletisation. They are also suitable for manual and automatic conveyor handling. The standard colour is grey.
Certifications: Guaranteed food-safe.
Ordering information: Trolleys and lids are optional and can be supplied for the entire range.
Product Family Options
Containers
Product Information
- 容量説明W×D×H ext.W×D×H int.AvailabilityPrice
- 2,9 LSolid Euro container, 15 kg load capacity, closed handles300×200×70 mm268×168×68 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2390 - 5,4 LSolid Euro container, 25 kg load capacity, closed handles400×300×70 mm367×268×58 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2392 - 7,2 LSolid Euro container, 15 kg load capacity, closed handles300×200×170 mm268×168×168 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2391 - 11,2 LSolid Euro container, 25 kg load capacity, closed handles400×300×120 mm367×268×117 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2393 - 13,7 LSolid Euro container, 25 kg load capacity, closed handles600×400×70 mm567×367×67 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2398 - 15,9 LSolid Euro container, 25 kg load capacity, open handles400×300×170 mm367×268×167 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2394 - 20,8 LSolid Euro container, 25 kg load capacity, open handles400×300×220 mm367×268×217 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2395 - 23,9 LSolid Euro container, 25 kg load capacity, closed handles600×400×120 mm567×367×117 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2399 - 25,6 LSolid Euro container, 25 kg load capacity, open handles400×300×270 mm367×268×267 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2396 - 30 LSolid Euro container, 25 kg load capacity, open handles400×300×320 mm367×268×317 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2397 - 35,6 LSolid Euro container, 25 kg load capacity, open handles600×400×170 mm567×367×167 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2400 - 44,2 LSolid Euro container, 25 kg load capacity, open handles600×400×220 mm567×367×217 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2401 - 54,4 LSolid Euro container, 25 kg load capacity, open handles600×400×270 mm567×367×267 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2402 - 64,5 LSolid Euro container, 25 kg load capacity, open handles600×400×320 mm567×267×317 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2409 - 91 LSolid Euro container, 35 kg load capacity, closed handles800×600×220 mm767×567×216 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2410 - 168 LSolid Euro container, 35 kg load capacity, closed handles800×600×420 mm767×567×416 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.216-2416
Lids
Product Information
- Catalog #説明寸法AvailabilityPrice