VWR®, LED蛍光キューブ
Supplier: VWR Collection
Jump to:
About this item
For use with microscope series IT406/407.
9 Options Available Below
Product Details & Documents
For use with microscope series IT406/407.
Documents
Product Family Options
Product Information
- 説明Optical featuresAvailabilityPrice
- Blue LED fluorescence cube for IT406/407LED emission 460 nm,
Excitation filter 455 - 495 nm,
Dichroic mirror cut-off 500 nm,
Emission filter 510 LP nmLog in for availability1 * 1 items - Green LED fluorescence cube for IT406/407LED emission 523 nm,
Excitation filter 510 -5 50 nm,
Dichroic mirror cut-off 570 nm,
Emission filter 575 LP nmLog in for availability1 * 1 items - Violet LED fluorescence cube for IT406/407LED emission 405 nm,
Excitation filter 390 - 420 nm,
Dichroic mirror cut-off 440 nm,
Emission filter 450 LP nmLog in for availability1 * 1 items - UV LED fluorescence cube for IT406/407LED emission 365 nm,
Excitation filter 325 - 375 nm,
Dichroic mirror cut-off 415 nm,
Emission filter 435 LP nmLog in for availability1 * 1 items - Red 1 LED fluorescence cube for IT406/407LED emission 623 nm,
Excitation filter 590 - 650 nm,
Dichroic mirror cut-off 660 nm,
Emission filter 665 LP nmLog in for availability1 * 1 items - Red 2 LED fluorescence cube for IT406/407LED emission 623 nm,
Excitation filter 595 - 645 nm,
Dichroic mirror cut-off 655 nm,
Emission filter 665 - 715 nmLog in for availability1 * 1 items - Deep red LED fluorescence cube for IT406/407LED emission 660 nm,
Excitation filter 623 - 678 nm,
Dichroic mirror cut-off 685 nm,
Emission filter 690 - 750 nmLog in for availability1 * 1 items - Far red LED fluorescence cube for IT406/407LED emission 740 nm,
Excitation filter 720 - 760 nm,
Dichroic mirror cut-off 770 nm,
Emission filter 780 LP nmLog in for availability1 * 1 items - Amber LED fluorescence cube for IT406/407LED emission 590 nm,
Excitation filter 582 - 603 nm,
Dichroic mirror cut-off 610 nm,
Emission filter 615 - 645 nmLog in for availability1 * 1 items