Orders LinkContactUsLinkComponent
VWR®, マルチチューブラック、ブルー/ナチュラル
VWR®, マルチチューブラック、ブルー/ナチュラル
Supplier:   VWR Collection
VWR®, マルチチューブラック、ブルー/ナチュラル
Supplier:  VWR Collection
Certificates
VWR®, マルチチューブラック、ブルー/ナチュラル
VWR®, マルチチューブラック、ブルー/ナチュラル
2 Options Available Below
Product Details & Documents
These flexible tube racks are designed for use with a multitude of tube diameters and heights. Each rack holds virtually any brand tube between 10 and 30 mm.
  • Rotate tube rack insert to an angle position to facilitate more comfortable pipetting
  • Rack wells have a unique teardrop design that enables better retention of tubes over traditional designs
  • Open design allows for better filling visibility of tube graduations when filling
  • Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used
  • Interchangeable tube rack inserts hold multiple tube sizes firmly in place when alternating between sample vessels
納品情報: Each rack includes 6 inserts (2 each/diameter).
Product Family Options
Product Information
  • 説明
    穴の直径
    穴の数
    For tubes
    W×D×H
    Availability
    Price
  • Blue/Natural
    13/17/30 mm
    20/20/6
    10 - 30 mm
    309×107×77 mm
    1 * 1 items
  • Purple/Natural
    13/17/30 mm
    20/20/6
    10 - 30 mm
    309×107×77 mm