Orders LinkContactUsLinkComponent
プレート、スクエアウェル、J.T.BAKER®
 :   Avantor
プレート、スクエアウェル、J.T.BAKER®
 :  Avantor
プレート、スクエアウェル、J.T.BAKER®
プレート、スクエアウェル、J.T.BAKER®
These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
  • Chemical resistant
  • V-bottom shape for 24- and 48-wells maximises liquid removal and aids resuspension
  • U-bottom shape for 96 wells
  • No inner edges to interfere with magnetic bead collection
  • Raised well rims to improve heat sealing
  • Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilisation.
  • Catalog No:
  • 43001-0020
  • 43001-0062
  • 43001-0066
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Product Family Options
  • 説明
    Colour
    Sterile
    Well volume
  • 96-well plate
    Transparent
    -
    2,0 ml
    1 * 20 items
    1 * 20 items - $153.50
    $153.50
    1 * 20 items
       $153.50
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 48-well plate
    Transparent
    -
    5,0 ml
    1 * 20 items
    1 * 20 items - $220.09
    $220.09
    1 * 20 items
       $220.09
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0062
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 24-well plate
    Transparent
    -
    10,0 ml
    1 * 20 items
    1 * 20 items - $227.58
    $227.58
    1 * 20 items
       $227.58
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0066
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Aluminium foil seals for long-term storage, chemical resistant (DMSO)
  •  
    説明
    厚さ
    Sterile
    光学特性
  • Sealing foil, large
    78,8 µm
    -
    Light blocking
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Accessories
  •  
    説明
    光学特性
    Material
  • 43001-0022
    Capmat for J.T.Baker plate square well 96
    Transparent
    EVA
    1 * 20 items - $114.50
  •  
    説明
    光学特性
    Material
  • 43001-0069
    Capmat for J.T.Baker plate square well 48
    Transparent
    EVA
    1 * 20 items - $185.32
  •  
    説明
    光学特性
    Material
  • 43001-0070
    Capmat for J.T.Baker plate square well 24
    Transparent
    EVA
    1 * 20 items - $125.39