Orders LinkContactUsLinkComponent
プレート、丸型ウェル、J.T.Baker®
プレート、丸型ウェル、J.T.Baker®
   Sustainable Options Available
Supplier:   Avantor
プレート、丸型ウェル、J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
プレート、丸型ウェル、J.T.Baker®
3 Options Available Below
Product Details & Documents
Made of virgin PP to minimise extractables.
  • Raised rim to prevent cross contamination and enable a better heat-welded seal
  • Also available rimless to allow insertion of vials, or when adhesive seals are used
  • Round bottom for optimal mixing and recovery
  • Suitable for use with automated sample handling systems
  • Can be stored at –80 °C
J.T.Baker Plate Round Well Plus with air escapes and raised rims for unfreezing samples.
Specifications for Product Family
Specifications
  • Catalog No:
  • 43001-0010
  • 43001-0016
  • 43001-0116
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Specifications
Product Family Options
Product Information
  • 説明
    Colour
    Sterile
    Well volume
    Availability
    Price
  • PS plates
    Clear
    -
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 20 items
    1 * 20 items - $174.44
    $174.44
    1 * 20 items
       $174.44
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0010
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PP plates
    Transparent
    -
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 20 items
    1 * 20 items - $174.55
    $174.55
    1 * 20 items
       $174.55
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0016
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PP plates plus, raised well rims
    Transparent
    -
    1,0 ml
    Log in for availability
    Log in for availability
    1 * 20 items
    1 * 20 items - $207.14
    $207.14
    1 * 20 items
       $207.14
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0116
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Aluminium foil seals for long-term storage, chemical resistant (DMSO)
Product Information
  • Catalog #
    説明
    厚さ
    Sterile
    光学特性
    Availability
    Price
  • Sealing foil, large
    78,8 µm
    -
    Light blocking
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00
    $0.00
    1 * 100 items
       $0.00
Accessories
Product Information
  • Catalog #
    説明
    光学特性
    Material
    Availability
    Price
  • 43001-0017
    Capmat for J.T.Baker Plate Round Well
    Transparent
    EVA
    Log in for availability
    1 * 20 items - $117.73