Orders LinkContactUsLinkComponent
J.T.Baker® プレートメディオ
   Sustainable Options Available
Supplier:   Avantor
J.T.Baker® プレートメディオ
Supplier:  Avantor
   Sustainable Options Available
Certificates
J.T.Baker® プレートメディオ
3 Options Available Below
Product Details & Documents
PP, 96-well storage plate with conical wells.
  • Due to the tangency of J.T.Baker Plate Medio plates, less than 50% stacking space is needed
  • Alphanumeric code to assist identification and registration of single samples
  • Higher well rims allow an airtight seal
  • Conical wells aid sample mixing
J.T.Baker Plate Medio is perfectly made for limited storage areas (fridge).
Specifications for Product Family
Specifications
  • Catalog No:
  • 43001-0102
  • 43001-0103
  • 43001-0101
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Specifications
Product Family Options
Product Information
  • Colour
    Recommended working volume
    Sterile
    Well volume
    Availability
    Price
  • Transparent
    20 - 500 µl
    -
    0,65 ml
    Log in for availability
    Log in for availability
    1 * 40 items
    1 * 40 items - $337.99
    $337.99
    1 * 40 items
       $337.99
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0102
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • Transparent
    50 - 2000 µl
    -
    2,3 ml
    Log in for availability
    Log in for availability
    1 * 20 items
    1 * 20 items - $157.53
    $157.53
    1 * 20 items
       $157.53
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0103
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • Transparent
    50 - 1000 µl
    -
    1,2 ml
    Log in for availability
    Log in for availability
    1 * 32 items
    1 * 32 items - $366.22
    $366.22
    1 * 32 items
       $366.22
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0101
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    説明
    光学特性
    Material
    Availability
    Price
  • 43001-0104
    Capmat for J.T.Baker® Plate Medio
    Transparent
    EVA
    Log in for availability
    1 * 20 items - $109.03