Orders LinkContactUsLinkComponent
384プレート、J.T.Baker®製
384プレート、J.T.Baker®製
   Sustainable Options Available
Supplier:   Avantor
384プレート、J.T.Baker®製
Supplier:  Avantor
   Sustainable Options Available
Certificates
384プレート、J.T.Baker®製
2 Options Available Below
Product Details & Documents
Available in PP for automated screening and storage systems.
  • Alphanumeric code to assist sample identification
  • Square wells for increased volume
  • V-bottom for high sample recovery
  • High frame (3 mm) for stable and slip-free stacking
J.T.Baker Plate 384 is ideal for small sample volumes.
Specifications for Product Family
Specifications
  • Catalog No:
  • 43004-0120
  • 43004-0200
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Specifications
Product Family Options
Product Information
  • 説明
    Colour
    Recommended working volume
    Sterile
    Well volume
    Availability
    Price
  • PP plates
    Transparent
    5 - 110 µl
    -
    120 µl
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $759.61
    $759.61
    1 * 60 items
       $759.61
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43004-0120
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PP plates
    Transparent
    5 - 180 µl
    -
    200 µl
    Log in for availability
    Log in for availability
    1 * 40 items
    1 * 40 items - $608.80
    $608.80
    1 * 40 items
       $608.80
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43004-0200
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Aluminium foil seals for long-term storage, chemical resistant (DMSO)
Product Information
  • Catalog #
    説明
    厚さ
    Sterile
    光学特性
    Availability
    Price
  • Sealing foil, large
    78,8 µm
    -
    Light blocking
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00