Orders LinkContactUsLinkComponent
J.T.Baker®プレート96
 :   Avantor
J.T.Baker®プレート96
 :  Avantor
J.T.Baker®プレート96
J.T.Baker®プレート96
J.T.Baker Plate 96 round well plates made from PS are ideal for ELISA applications.
  • Alphanumeric coding assists with identification of samples
  • Plate shape significantly reduces sample carry-over
  • Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
  • Catalog No:
  • 43001-0117
  • 43001-0118
  • 43001-0119
  • 43001-0600
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
Product Family Options
  • 説明
    Colour
    Sterile
    Well volume
  • PS plates, U-bottom
    Clear
    -
    0,3 ml
    1 * 100 items
    1 * 100 items - $278.00
    $278.00
    1 * 100 items
       $278.00
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0117
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PS plates, V-bottom
    Clear
    -
    0,24 ml
    1 * 100 items
    1 * 100 items - $261.65
    $261.65
    1 * 100 items
       $261.65
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0118
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PS plates, F-bottom
    Clear
    -
    0,35 ml
    1 * 100 items
    1 * 100 items - $268.38
    $268.38
    1 * 100 items
       $268.38
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0119
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • PP plates, U-bottom
    Transparent
    -
    0,3 ml
    1 * 64 items
    1 * 64 items - $348.32
    $348.32
    1 * 64 items
       $348.32
      Low Manufacturing Impact
    Specifications
    Cat. No.
    43001-0600
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
      Low Manufacturing Impact