Orders LinkContactUsLinkComponent
J.T.Baker® プレート SRW、マグネティック
J.T.Baker® プレート SRW、マグネティック
   Sustainable Options Available
Supplier:   Avantor
J.T.Baker® プレート SRW、マグネティック
Supplier:  Avantor
   Sustainable Options Available
Certificates
J.T.Baker® プレート SRW、マグネティック
J.T.Baker® プレート SRW、マグネティック
6 Options Available Below
Product Details & Documents
These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as the KingFisher™ Apex, Flex 96 and Presto.
  • Standard SBS footprint
  • Cylindrical wells enable good sample mixing
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code assists registration and identification of individual samples
Specifications for Product Family
Specifications
  • Catalog No:
  • 43001-0506
  • 43001-0514
  • 43001-0516
  • 43001-0505
  • 43001-0504
  • 43001-0515
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
  • ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
Specifications
Product Family Options
Plates
Product Information
  • 説明
    Colour
    Well volume
    Sterile
    Availability
    Price
  • 96-well plate, square wells, V-bottom
    Transparent
    0,2 ml
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $508.09
    $508.09
    1 * 60 items
       $508.09
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0506
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 96-well plate, square wells, V-bottom, with barcode
    Transparent
    2,0 ml
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $618.21
    $618.21
    1 * 60 items
       $618.21
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0514
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
      Low Manufacturing Impact
  • 96-well plate, square wells, V-bottom, with barcode
    Transparent (Removed)
    0,2 ml
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $549.51
    $549.51
    1 * 60 items
       $549.51
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0516
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
      Low Manufacturing Impact
  • 96-well plate, square wells, V-bottom
    Transparent
    2,0 ml
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $613.19
    $613.19
    1 * 60 items
       $613.19
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0504
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Tip comb
Product Information
  • 説明
    Colour
    Well volume
    Sterile
    Availability
    Price
  • Tip comb
    Transparent
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $655.48
    $655.48
    1 * 60 items
       $655.48
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0505
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • Tip comb with barcode
    Transparent (Removed)
    -
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - $738.38
    $738.38
    1 * 60 items
       $738.38
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    43001-0515
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
      Low Manufacturing Impact
Aluminium foil seals for long-term storage, chemical resistant (DMSO)
Product Information
  • Catalog #
    説明
    厚さ
    Sterile
    光学特性
    Availability
    Price
  • Sealing foil, large
    78,8 µm
    -
    Light blocking
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - $0.00