J.T.Baker®ロボットチップMP型
1000 µl, -, +, Black, 8× 5 racks, 96 each, blistered
Catalog # 49209-0105
Supplier: Avantor
Jump to:
Product Details & Documents
Suitable for use with Roche MagNA Pure instruments. Available in conductive version, with filter, and in CE-IVD marked version for clinical diagnostic applications.
- Manufactured under cleanroom conditions (cleanroom Class 8)
- State-of-the-art quality control
- Barcoded trays for automated identification
- Free of DNA, DNase, RNase and PCR inhibitors
- Ideal for high throughput analysis
- Packed in blisters
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Specifications
- Packed:8× 5 racks, 96 each, blistered
- Sterile:-
- 容量:1000 µl
- Seq:02
- Tip colour:Black
- Filter:+
- Cat. No.:49209-0105
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact