Orders LinkContactUsLinkComponent
J.T.Baker®ロボットチップDX型
 :   Avantor
J.T.Baker®ロボットチップDX型
 :  Avantor
J.T.Baker®ロボットチップDX型
J.T.Baker®ロボットチップDX型
Suitable for use on Dynex DSX and DS2 instruments. Available in non conductive version, without filter, in container rack, and in CE-IVD marked version for clinical diagnostic applications.
  • 108 tips in one optimized container with lid
  • Especially developed for the automated sample handling
  • Highest quality specifications for the compatibility and cleanliness
  • Blue sample tips for visual tip identification
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
  • Catalog No:
  • 49136-0000
  • 49137-0000
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Product Family Options
  • 容量
    Sterile
    Tip colour
    Packed
  • 300 µl
    -
    Blue
    1× 4 racks, 108 each
    1 * 432 items
    1 * 432 items - $111.26
    $111.26
    1 * 432 items
       $111.26
      Low Manufacturing Impact
    Specifications
    Cat. No.
    49136-0000
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
  • 1300 µl
    -
    Clear
    1× 4 racks, 108 each
    1 * 432 items
    1 * 432 items - $109.49
    $109.49
    1 * 432 items
       $109.49
      Low Manufacturing Impact
    Specifications
    Cat. No.
    49137-0000
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
      Low Manufacturing Impact
Accessories
  •  
    Colour
    Sterile
  • 43001-0031
    Transparent
    -
    1 * 250 items - $272.59