J.T.Baker®ロボットチップQG型
50 µl, -, +, Black, 12× 2 racks, 96 each, blistered
Catalog # 49018-0106
Supplier: Avantor
Jump to:
Product Details & Documents
Suitable for use on third party robotic instruments QIAgility. Available in conductive version, with filter, non-sterile, and in CE-IVD marked version for clinical diagnostic applications.
- Manufactured under cleanroom conditions (cleanroom class 8)
- State of the art quality control
- Free of DNA, DNase, RNase and PCR inhibitors
- Ideal for high-throughput analysis and DNA purification
- Packed in blister
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Documents
Specifications
- Packed:12× 2 racks, 96 each, blistered
- Sterile:-
- 容量:50 µl
- Seq:1
- Tip colour:Black
- Filter:+
- Cat. No.:49018-0106
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact