Orders LinkContactUsLinkComponent
VWR®, 精密グリップグローブ、PUコーティング
VWR®, 精密グリップグローブ、PUコーティング
Supplier:   VWR Collection
VWR®, 精密グリップグローブ、PUコーティング
Supplier:  VWR Collection
Certificates
VWR®, 精密グリップグローブ、PUコーティング
VWR®, 精密グリップグローブ、PUコーティング
6 Options Available Below
Product Details & Documents
VWR® precision grip gloves with polyurethane coating provide basic cut protection and protection against mechanical risks.
  • ANSI A1 cut protection (Level A)
  • Nylon composition
  • Polyurethane (PU) coating on palm and fingertips
  • Black knitted glove with black coating
Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment. Also great for package handling, picking and packing.
Certifications: Meets requirements for ANSI A1 Cut Resistance. EN 388:2016 (4131X)
Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-5697
  • 112-5698
  • 112-5699
  • 112-5726
  • 112-5727
  • 112-5728
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Finish:
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • PU palm coating
  • Gauge:
  • 15
  • 15
  • 15
  • 15
  • 15
  • 15
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4131X)
  • EN 388:2016 (4131X)
  • EN 388:2016 (4131X)
  • EN 388:2016 (4131X)
  • EN 388:2016 (4131X)
  • EN 388:2016 (4131X)
Specifications
Product Family Options
Product Information
  • 手袋のサイズ
    Colour
    Availability
    Price
  • 6
    Black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5697
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4131X)
  • 7
    Black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5698
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4131X)
  • 8
    Black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5699
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4131X)
  • 9
    Black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5726
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4131X)
  • 10
    Black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5727
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4131X)
  • 11
    Black
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    112-5728
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex / latex-free
    Latex-free
    Finish
    PU palm coating
    Gauge
    15
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4131X)